Skip to Content
Merck
All Photos(4)

Key Documents

L4533

Sigma-Aldrich

Lithium hydroxide monohydrate

BioXtra, 98.5-101.5% (titration)

Synonym(s):

LiOH, LiOH.H2O, Lithium Hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiOH · H2O
CAS Number:
Molecular Weight:
41.96
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21

product line

BioXtra

Quality Level

Assay

98.5-101.5% (titration)

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

solubility

H2O: 1 M, clear to slightly hazy, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.01%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[Li+].O.[OH-]

InChI

1S/Li.2H2O/h;2*1H2/q+1;;/p-1

InChI key

GLXDVVHUTZTUQK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Deliquescent Lithium Sulfide Synthesis: Research on the air synthesis of deliquescent lithium sulfide demonstrates the versatility of lithium-based compounds, including lithium hydroxide monohydrate, in developing new materials for energy storage and battery technology (Yang et al., 2023).

  • Artificial Solid Electrolyte Interphase: Lithium hydroxide monohydrate′s potential utility in stabilizing lithium metal anodes at high rates of operation is indicated by studies on catalytic chemistry derived artificial solid electrolyte interphase, enhancing the safety and efficacy of lithium-ion batteries (Cheng et al., 2023).

  • Synthesis of Functionalized Disiloxanes: The study involving the synthesis of sulfur-containing functionalized disiloxanes showcases the broad applicability of innovative synthetic methods which could also include lithium hydroxide monohydrate to create materials with nonconventional properties, relevant in various chemical and industrial processes (Tang et al., 2023).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayorinde Awonusi et al.
Calcified tissue international, 81(1), 46-52 (2007-06-07)
A series of apatites with varying carbonate levels was prepared in order to assign the carbonate bands and calibrate for Raman analysis of natural materials. Overlap of carbonate bands with phosphate peaks was resolved by curve fitting. A peak at
L C Hsu et al.
Journal of hazardous materials, 142(1-2), 242-249 (2006-09-19)
Hexavalent Cr has been identified as one of the toxic metals commonly present in industrial effluents. Among the treatment techniques developed for removing Cr(VI) from waste waters, sorption is most commonly applied, due to its simplicity and efficiency. However, few
Anupriya Veerman et al.
The Journal of chemical physics, 123(8), 084321-084321 (2005-09-17)
The structures, stabilities, thermodynamic quantities, dissociation energies, infrared spectra, and electronic properties of LiOH hydrated by up to seven water molecules are investigated by using the density-functional theory and the Møller-Plesset second-order perturbation theory (MP2). Further accurate analysis based on
Yuan Gee Lee et al.
Biosensors & bioelectronics, 20(1), 33-40 (2004-05-15)
Ionic liquids containing lithium methylsulfonyl group were prepared from the precursors poly(propylene glycol)-block-(ethylene glycol)-block-(propylene glycol)-bis(2-aminopropyl ether) with different molecular weight. These liquids revealed excellent electrical conductivity in the temperature range -25 to 85 degrees C. Also, they exhibited a high
Kwang-Heon Kim et al.
Ultrasonics sonochemistry, 15(6), 1019-1025 (2008-05-09)
Nano-sized HT-LiCoO(2) powders were prepared by sonochemical synthesis in an aqueous solution of lithium hydroxide containing cobalt hydroxide at approximately 80 degrees C without any further heat treatment at high temperature. The effects of the LiOH concentration, oxidation conditions, ultrasound

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service