Skip to Content
Merck
All Photos(1)

Key Documents

IRMM316

Nivalenol in acetonitrile

IRMM®, certified reference material

Synonym(s):

Nivalenol solution, 3α,4β,7α, 15-Tetrahydroxy-12,13-epoxytrichothec-9-en-8-one, NIV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O7
CAS Number:
Molecular Weight:
312.32
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

manufacturer/tradename

JRC

application(s)

general analytical

format

matrix material

SMILES string

[H][C@]12O[C@]3([H])[C@H](O)[C@@H](O)[C@@](C)([C@]34CO4)[C@@]1(CO)[C@H](O)C(=O)C(C)=C2

InChI

1S/C15H20O7/c1-6-3-7-14(4-16,11(20)8(6)17)13(2)10(19)9(18)12(22-7)15(13)5-21-15/h3,7,9-12,16,18-20H,4-5H2,1-2H3/t7-,9-,10-,11-,12-,13-,14-,15+/m1/s1

InChI key

UKOTXHQERFPCBU-XBXCNEFVSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
IRMM316

Legal Information

IRMM is a registered trademark of European Commission

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Barros et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 29(2), 293-303 (2011-05-21)
Soybean (Glycine max L.), the main source of protein throughout the world, is used both as a food and a feedstuff. Currently, limited information about the occurrence of Fusarium species and mycotoxins in soybean grain and by-products is available. The
E D Van Asselt et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 29(10), 1556-1565 (2012-06-30)
Mycotoxins are secondary metabolites produced by fungi that can cause adverse health effects. Due to climate change, temperatures are expected to rise and changes in rainfall patterns are foreseen. These developments may increase fungal occurrence and mycotoxin concentrations in maize.
Theresa Lee et al.
Journal of food protection, 74(7), 1169-1174 (2011-07-12)
A total of 201 samples of brown rice, polished rice, and two types of by-products, blue-tinged rice and discolored rice, were collected from rice stores maintained at 51 rice processing complexes in Korea. These samples were analyzed for the presence
Jungkwan Lee et al.
Applied and environmental microbiology, 78(7), 2161-2167 (2012-01-31)
Fusarium graminearum (Gibberella zeae) is an important pathogen of wheat, maize, barley, and rice in South Korea, and harvested grain often is contaminated with trichothecenes such as deoxynivalenol and nivalenol. In this study, we examined 568 isolates of F. graminearum
Marilena Muscarella et al.
Talanta, 97, 145-149 (2012-07-31)
A rapid, sensitive and selective analytical method was developed for the quantitative determination of deoxynivalenol (DON) and nivalenol (NIV) in cereals intended for human and animal consumption. The method, based on liquid chromatography and fluorescence detection, involves an automated 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service