Skip to Content
Merck
All Photos(1)

Key Documents

I0380000

Isoconazole nitrate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14Cl4N2O · HNO3
CAS Number:
Molecular Weight:
479.14
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

isoconazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1c(c(ccc1)Cl)COC(C[n]3cncc3)c2c(cc(cc2)Cl)Cl.[N+](=O)([O-])O

InChI

1S/C18H14Cl4N2O.HNO3/c19-12-4-5-13(17(22)8-12)18(9-24-7-6-23-11-24)25-10-14-15(20)2-1-3-16(14)21;2-1(3)4/h1-8,11,18H,9-10H2;(H,2,3,4)

InChI key

NNGQLSIGRSTLLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isoconazole nitrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcus Henrique de S B Xavier et al.
Dermatology online journal, 14(8), 15-15 (2008-12-09)
Tinea nigra is an asymptomatic superficial fungal infection caused by Phaeoannelomyces werneckii, generally affecting the skin of the palms and characterized by deeply pigmented macular non-scaly patches. These lesions are quite characteristic. However, they can be misdiagnosed as a malignant
J Lademann et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 80(3), 615-620 (2012-01-31)
Bacteria and fungi are located in the stratum corneum and the hair follicles. Therefore, the development and assessment of efficient drugs requires standard in vivo investigation methods permitting a differentiation between intercellular and follicular penetration and storage of topically applied
Tinea cruris treated with a combined topical therapy containing isoconazole and a corticosteroid.
Jaco Jürgens
Mycoses, 51 Suppl 4, 32-33 (2008-09-12)
Irina Vennewald et al.
Clinics in dermatology, 28(2), 202-211 (2010-03-30)
Aspergillus and Candida spp are the most frequently isolated fungi in patients with otomycosis. The diagnosis of otitis externa relies on the patient's history, otoscopic examination under microscopic control, and imaging studies. Direct preparation of the specimens, particularly with optical
Tinea corporis of the shin and chest successfully treated with a topical antifungal and corticosteroid cream.
Ga-Young Lee et al.
Mycoses, 51 Suppl 4, 34-36 (2008-09-12)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service