F171503
Diamond 200 μL tip pack
for use with BioControl® GDS, suitable for PCR, tip volume 200 μL
Synonym(s):
Diamond 200 μL tip pack, Sterile Pipette Tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106302
NACRES:
NB.02
sterility
sterile
packaging
box of 10 × 96 units
technique(s)
PCR: suitable
tip volume
200 μL
compatibility
for use with BioControl® GDS
storage temp.
room temp
General description
Disposable tips are used to transfer PCR reagents. Can be used as consumable accessories during GDS PCR sample preparation.
Application
The Assurance® GDS Diamond Tips are used to transfer liquids during Assurance® GDS sample preparation.
Legal Information
Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
BioControl is a registered trademark of BioControl Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service