Skip to Content
Merck
All Photos(1)

Key Documents

E0109000

Enalapril maleate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Enalapril maleate salt, (S)-N-[1-(Ethoxycarbonyl)-3-phenylpropyl]-Ala-Pro maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28N2O5 · C4H4O4
CAS Number:
Molecular Weight:
492.52
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

enalapril

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H](C(N1[C@H](C(O)=O)CCC1)=O)N[C@H](CC(OCC)=O)CCC2=CC=CC=C2.O=C(OO)/C=C\C(O)=O

InChI

1S/C21H30N2O5.C4H4O5/c1-3-28-19(24)14-17(12-11-16-8-5-4-6-9-16)22-15(2)20(25)23-13-7-10-18(23)21(26)27;5-3(6)1-2-4(7)9-8/h4-6,8-9,15,17-18,22H,3,7,10-14H2,1-2H3,(H,26,27);1-2,8H,(H,5,6)/b;2-1-/t15-,17-,18-;/m0./s1

InChI key

UQKQUZOIQFOJOI-KJHLASMFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Enalapril maleate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

A long-acting angiotensin-converting enzyme inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Marta de Diego et al.
Journal of chromatographic science, 52(10), 1193-1197 (2013-12-04)
A simple and rapid stability-indicating liquid chromatographic method was developed and validated for the simultaneous determination of lisinopril and hydrochlorotiazide (HCTZ) in drug substances and dosage forms in the presence of degradation products. Forced degradation studies were conducted on the
Jorge Luiz Alves-Pereira et al.
The Journal of urology, 192(6), 1878-1883 (2014-06-01)
We evaluated whether antihypertensive drugs that act through the renin-angiotensin system would affect testis function. Ten mice were fed standard chow and 40 received a high energy density diet. At 8 weeks the high energy density diet mice were divided
A Uzawa et al.
Clinical and experimental immunology, 178(2), 245-252 (2014-07-06)
Multiple sclerosis (MS) is a chronic inflammatory disease of the central nervous system (CNS). Bradykinin is the end-product of the kallikrein/kinin system, which has been recognized as an endogenous target for combating CNS inflammation. Angiotensin-converting enzyme (ACE) inhibitors influence the
Bjoern B Burckhardt et al.
Biomedical chromatography : BMC, 28(12), 1679-1691 (2014-05-03)
The benefit-risk ratio of combined blocking by the direct renin inhibitor aliskiren and an angiotensin-converting enzyme inhibitor (e.g. enalapril) on the renin-angiotensin-aldosterone system is discussed. No method was available for simultaneous determination of both drugs in urine. A novel sensitive
John J V McMurray et al.
The New England journal of medicine, 371(11), 993-1004 (2014-09-02)
We compared the angiotensin receptor-neprilysin inhibitor LCZ696 with enalapril in patients who had heart failure with a reduced ejection fraction. In previous studies, enalapril improved survival in such patients. In this double-blind trial, we randomly assigned 8442 patients with class

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service