Skip to Content
Merck
All Photos(1)

Key Documents

D8536

Supelco

Diperodon hydrochloride

analytical standard

Synonym(s):

3-(1-Piperidinyl)-1,2-propanediol bis(phenylcarbamate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27N3O4 · HCl
CAS Number:
Molecular Weight:
433.93
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl.O=C(Nc1ccccc1)OCC(CN2CCCCC2)OC(=O)Nc3ccccc3

InChI

1S/C22H27N3O4.ClH/c26-21(23-18-10-4-1-5-11-18)28-17-20(16-25-14-8-3-9-15-25)29-22(27)24-19-12-6-2-7-13-19;/h1-2,4-7,10-13,20H,3,8-9,14-17H2,(H,23,26)(H,24,27);1H

InChI key

OWULVAZDMWJBLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Cizmárik et al.
Acta poloniae pharmaceutica, 53(3), 167-169 (1996-05-01)
A local anaesthetics diperodon by the action of hydrolazes in blood serum is decomposed. A method for its determination in blood serum in vitro has been worked out. It was found that during the first 2 hours it decomposes rather
K Hrobonová et al.
Ceska a Slovenska farmacie : casopis Ceske farmaceuticke spolecnosti a Slovenske farmaceuticke spolecnosti, 53(5), 225-227 (2004-10-28)
The chiral stationary phase on the base of teicoplanin and the polar-organic mobile phase methanol/acetonitrile/acetic acid/triethylamine 45/55/0.3/0.2 was used for the separation of diperodon enantiomers. The developed method was suitable to determine the enantiomers in blood serum up to 0.5
K Hrobonová et al.
Journal of pharmaceutical and biomedical analysis, 30(3), 875-880 (2002-10-09)
An on-line coupled HPLC system is described for the determination of the enantiomers of diperodon in blood serum. The method involves three steps: (i) off-line preconcentration and clean-up, (ii) separation of the diperodon enantiomers from the matrix components on a
Allergy to the local anaesthetic diperodon.
C D Calnan
Contact dermatitis, 6(5), 367-367 (1980-08-01)
Fei Wang et al.
Developmental cell, 52(2), 141-151 (2020-01-29)
We explored the potential for autophagy to regulate budding yeast meiosis. Following pre-meiotic DNA replication, we blocked autophagy by chemical inhibition of Atg1 kinase or engineered degradation of Atg14 and observed homologous chromosome segregation followed by sister chromatid separation; cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service