Skip to Content
Merck
All Photos(1)

Key Documents

D7439

Supelco

Dihydroartemisinin

analytical standard, mixture of α and β isomers

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O5
Molecular Weight:
284.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97% (TLC)

form

powder

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@H]1C(O)O[C@@]2([H])[C@@]3(OO4)[C@](CC[C@]4(C)O2)([H])[C@H](C)CC[C@]31[H]

InChI

1S/C15H24O5/c1-8-4-5-10-9(2)12(16)18-13-15(10)11(8)6-14(3,7-17-13)19-20-15/h8-13,16H,4-7H2,1-3H3/t8-,9-,10+,11+,12?,13+,14+,15+/m1/s1

InChI key

DJGUTYJDEZGRGH-NJEKVMANSA-N

General description

Antimalarial; antipsoriasis; anti-inflammatory.
Dihydroartemisinin is the major metabolite of the antimalarial agent, artemether. It is highly efficient against both drug-sensitive and drug-resistant strains of Plasmodium falciparum.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Warning

Protect from light.

Preparation Note

artemisia plant extract

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Org. Perox. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Marijon et al.
Malaria journal, 13, 501-501 (2014-12-18)
Improving management of patients suffering from cerebral malaria is needed to reduce the devastating mortality and morbidity of the disease in endemic areas. Intravenous artesunate is currently the first-line treatment, but the lack of material and skills in the field
Zenglei Wang et al.
PloS one, 9(4), e93825-e93825 (2014-04-17)
Malaria elimination/eradication campaigns emphasize interruption of parasite transmission as a priority strategy. Screening for new drugs and vaccines against gametocytes is therefore urgently needed. However, current methods for sexual stage drug assays, usually performed by counting or via fluorescent markers
Selective determination, in plasma, of artemether and its major metabolite, dihydroartemisinin, by high-performance liquid chromatography with ultraviolet detection
Thomas GC, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 583(1), 131-136 (1992)
Aditya S Paul et al.
Nature communications, 11(1), 3532-3532 (2020-07-17)
Asexual proliferation of the Plasmodium parasites that cause malaria follows a developmental program that alternates non-canonical intraerythrocytic replication with dissemination to new host cells. We carried out a functional analysis of the Plasmodium falciparum homolog of Protein Phosphatase 1 (PfPP1)
Lisa Evans DeWald et al.
Scientific reports, 9(1), 20199-20199 (2019-12-29)
During the Ebola virus disease (EVD) epidemic in Western Africa (2013‒2016), antimalarial treatment was administered to EVD patients due to the high coexisting malaria burden in accordance with World Health Organization guidelines. In an Ebola treatment center in Liberia, EVD patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service