Skip to Content
Merck
All Photos(1)

Key Documents

D2953000

Dithranol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,8,9-Anthracenetriol, 1,8-Dihydroxy-9(10H)-anthracenone, 1,8-Dihydroxyanthrone, Anthralin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O3
CAS Number:
Molecular Weight:
226.23
Beilstein:
2054360
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

anthralin, dithranol

manufacturer/tradename

EDQM

mp

178-181 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Oc1cccc2Cc3cccc(O)c3C(=O)c12

InChI

1S/C14H10O3/c15-10-5-1-3-8-7-9-4-2-6-11(16)13(9)14(17)12(8)10/h1-6,15-16H,7H2

InChI key

NUZWLKWWNNJHPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dithranol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yann Bernhard et al.
Dalton transactions (Cambridge, England : 2003), 44(7), 3200-3208 (2015-01-13)
Subphthalocyanine (SubPc), a putative fluorophore for optical imaging (OI), was conjugated to chelating ligands (DOTA, DTPA) affording water-soluble conjugates complexed with (non-radioactive) metals relevant to the following medical imaging techniques/therapies: MRI (Gd), PET (Cu, Ga), SPECT (In, Ga, Lu), RIT
Emilie Cossoul et al.
Analytica chimica acta, 856, 46-53 (2014-12-30)
Recently, the interest of the coupling between atmospheric solid analysis probe (ASAP) and ion mobility-mass spectrometry has been revealed in the field of polymers. This method associates a direct ionization technique with a bi-dimensional separation method. Poly(ether ether ketones) (PEEK)
Kuan Sun et al.
Nature communications, 6, 6013-6013 (2015-01-15)
Solution-processed organic photovoltaic cells (OPVs) hold great promise to enable roll-to-roll printing of environmentally friendly, mechanically flexible and cost-effective photovoltaic devices. Nevertheless, many high-performing systems show best power conversion efficiencies (PCEs) with a thin active layer (thickness is ~100 nm) that
Sanjib Banerjee et al.
Journal of chromatography. A, 1376, 98-104 (2014-12-24)
The chemical homogeneity of telechelic polyisobutylene diol (PIB-diol), prepared by hydroboration-oxidation of allyl telechelic PIB obtained by reacting living PIB with allyltrimethylsilane, was investigated by liquid chromatography at critical conditions (LCCC) and HPLC coupled with matrix-assisted laser desorption/ionization time-of-flight mass
Thierry Fouquet et al.
Analytica chimica acta, 843, 46-58 (2014-08-26)
Cardanol and cardanol derivatives are among the most important biobased materials currently investigated in green chemistry, as renewable and promising building blocks in lieu of traditional raw materials from non renewable resources, in particular owing to the olefinic linkages on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service