Skip to Content
Merck
All Photos(1)

Key Documents

D0125000

Daunorubicin hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Daunomycin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29NO10 · HCl
CAS Number:
Molecular Weight:
563.98
Beilstein:
4229221
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

daunorubicin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O[C@H]5C[C@H](N)[C@H](O)[C@H](C)O5)c4c(O)c3C(=O)c12)C(C)=O

InChI

1S/C27H29NO10.ClH/c1-10-22(30)14(28)7-17(37-10)38-16-9-27(35,11(2)29)8-13-19(16)26(34)21-20(24(13)32)23(31)12-5-4-6-15(36-3)18(12)25(21)33;/h4-6,10,14,16-17,22,30,32,34-35H,7-9,28H2,1-3H3;1H/t10-,14-,16-,17-,22+,27-;/m0./s1

InChI key

GUGHGUXZJWAIAS-QQYBVWGSSA-N

Gene Information

human ... TOP2A(7153)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Daunorubicin hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Potent anticancer agent. Inhibits DNA and RNA synthesis as sequence specific ds-DNA intercalating agent.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I-Shan Hsieh et al.
Molecular pharmacology, 83(5), 968-977 (2013-02-26)
Multidrug resistance is a major cause of chemotherapy failure. Recent studies indicate that drug resistance can be rapidly induced by some soluble factors, such as cytokines, chemokines, growth factors, and cell adhesion factors in the tumor microenvironment. Osteopontin (OPN), an
Jay Chhablani et al.
Investigative ophthalmology & visual science, 54(2), 1268-1279 (2013-01-17)
To test the feasibility of covalent loading of daunorubicin into oxidized porous silicon (OPS) and to evaluate the ocular properties of sustained delivery of daunorubicin in this system. Porous silicon was heat oxidized and chemically functionalized so that the functional
Cedric Dos Santos et al.
Blood, 122(11), 1900-1913 (2013-07-31)
The SRC family kinases (SFKs) and the receptor tyrosine kinase c-Kit are activated in human acute myeloid leukemia (AML) cells. We show here that the SFKs LYN, HCK, or FGR are overexpressed and activated in AML progenitor cells. Treatment with
Sarah Bertoli et al.
Blood, 121(14), 2618-2626 (2013-02-01)
In acute myeloid leukemia (AML), new strategies assess the potential benefit of genetically targeted therapy at diagnosis. This implies waiting for laboratory tests and therefore a delay in initiation of chemotherapy. We studied the impact of time from diagnosis to
Mona Hassanein et al.
Leukemia research, 37(5), 556-560 (2013-01-30)
We evaluated outcomes in two consecutive groups of AML patients age>60 years in CR after 7+3 induction therapy. Group 1 received consolidation with cytarabine 1.5g/m(2) q12h×6+daunorubicin for two cycles, while group 2 received consolidation with 7+3 followed by mitoxantrone+etoposide. For

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service