CRM046
Trace Metals - Clay 1
certified reference material, pkg of 50 g
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
CofA
current certificate can be downloaded
packaging
pkg of 50 g
manufacturer/tradename
RTC CRM046-050
technique(s)
AAS: suitable
ICP: suitable
pH
5.39
application(s)
environmental
format
matrix material
storage temp.
2-30°C
Related Categories
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
This product was produced and certified from a naturally contaminated matrix and has not been fortified or modified in any way. Analytical data for Certification was obtained using USEPA SW846, 3rd edition methods 3050 (hot block) and 3051(microwave) using nitric acid extraction. Analysis was carried out according to USEPA methods 6010 (ICP-ES), 6020 (ICP-MS) and 7000 (AES).
Other Notes
Values of analytes vary lot to lot. Some lots may not contain all analytes.
Disclaimer
For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.


Analyte
Description
Arsenic
Cadmium
Chromium
Cobalt
Copper
Lead
Manganese
Mercury
Nickel
Zinc
See All (10)
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Skin Sens. 1
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service