Skip to Content
Merck
All Photos(1)

Key Documents

C1905000

Chlorpropamide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-(p-Chlorobenzenesulfonyl)-3-propylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13ClN2O3S
CAS Number:
Molecular Weight:
276.74
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

chlorpropamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCNC(=O)NS(=O)(=O)c1ccc(Cl)cc1

InChI

1S/C10H13ClN2O3S/c1-2-7-12-10(14)13-17(15,16)9-5-3-8(11)4-6-9/h3-6H,2,7H2,1H3,(H2,12,13,14)

InChI key

RKWGIWYCVPQPMF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chlorpropamide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Flush induced by chlorpropamide-alcohol (CAF): a genetic marker of diabetes mellitus or an undesirable effect of the drug?].
J Sabán et al.
Revista clinica espanola, 176(3), 107-111 (1985-02-01)
The chlorpropamide alcohol flush.
P G Wiles et al.
Clinical science (London, England : 1979), 67(4), 375-381 (1984-10-01)
Jing Li et al.
Critical care medicine, 42(12), e734-e740 (2014-11-18)
Cooling following cardiac arrest can improve survival significantly. However, delays in achieving target temperature may decrease the overall benefits of cooling. Here, we test whether lipid emulsion, a clinically approved drug reported to exert cardioprotection, can rescue heart contractility in
[Chlorpropamide-alcohol test as a marker of vascular complications of non-insulin-dependent diabetes mellitus].
M I Balabolkin et al.
Terapevticheskii arkhiv, 56(12), 128-130 (1984-01-01)
A hypothesis for the molecular mechanism of action of chlorpropamide in the treatment of diabetes mellitus and diabetes insipidus.
R M Zusman et al.
Federation proceedings, 36(13), 2728-2729 (1977-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service