Skip to Content
Merck
All Photos(1)

Key Documents

BP464

Prednisolone

British Pharmacopoeia (BP) Assay Standard

Synonym(s):

1,4-Pregnadiene-11β,17α,21-triol-3,20-dione, 1-Dehydrocortisol, 1-Dehydrohydrocortisone, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O5
CAS Number:
Molecular Weight:
360.44
Beilstein:
1354103
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

prednisolone

form

crystalline

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

240 °C (dec.) (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

SMILES string

O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(C)[C@@]3([H])CC[C@]4(O)C(CO)=O)=C1

InChI

1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-16,18,22,24,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

OIGNJSKKLXVSLS-VWUMJDOOSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Prednisolone BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Estriol Cream
  • Prednisolone Sodium Phosphate Ear Drops
  • Prednisolone Sodium Phosphate Eye Drops
  • Prednisolone Tablets
  • Prednisolone Soluble Tablets
  • Prednisolone Enema
  • Prednisolone Sodium Phosphate Oral Solution
  • Prednisolone Gastro-resistant Tablets
  • Other Notes

    Sales restrictions may apply.

    related product

    Product No.
    Description
    Pricing

    Pictograms

    Health hazard

    Signal Word

    Danger

    Hazard Statements

    Hazard Classifications

    Repr. 1B

    Storage Class Code

    6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

    WGK

    WGK 2


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Eng Cern Gan et al.
    International forum of allergy & rhinology, 4(11), 877-884 (2014-08-20)
    The endoscopically magnified operative field in functional endoscopic sinus surgery (FESS) makes even a small amount of bleeding a potentially significant hindrance. It is thought that irrigation with hot saline during surgery may improve surgical field of view by producing
    Ramachandran Venkitaraman et al.
    European urology, 67(4), 673-679 (2014-12-03)
    Prednisolone is widely used as secondary hormonal treatment for castration-resistant prostate cancer (CRPC). We hypothesised that dexamethasone, another corticosteroid, is more active. To compare the activity of prednisolone and dexamethasone in CRPC. This single-centre, randomised, phase 2 trial was performed
    T Bakchoul et al.
    Journal of thrombosis and haemostasis : JTH, 12(6), 871-878 (2014-03-25)
    Heparin, the standard perioperative anticoagulant for the prevention of graft vessel thrombosis in patients undergoing liver transplantation (LT), binds to the chemokine platelet factor 4 (PF4). Antibodies that are formed against the resulting PF4/heparin complexes can induce heparin-induced thrombocytopenia. LT
    C Motteram et al.
    Reproductive biomedicine online, 30(4), 340-348 (2015-02-14)
    The effectiveness of combined co-treatment with aspirin, doxycycline, prednisolone, with or without oestradiol patches, was investigated on live birth (LBR) rates after fresh and frozen embryo transfers (FET) in IVF and intracytoplasmic sperm injection cycles. Cases (n = 485) and
    M Tyblova et al.
    European journal of neurology, 22(4), 624-632 (2014-06-17)
    The prevalence of osteopenia and osteoporosis is higher amongst patients with multiple sclerosis in comparison with the general population. In addition to the general determinants of bone health, two factors may contribute to reduced bone mineral density in multiple sclerosis:

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service