BCR479
Fresh water (nitrate, low level)
BCR®, certified reference material
About This Item
Recommended Products
grade
certified reference material
Agency
BCR®
manufacturer/tradename
JRC
format
matrix material
1 of 4
This Item | WHA68097003 | WHA68097023 | WHA68095012 |
---|---|---|---|
0.1 μm | 0.02 μm | 0.2 μm | 0.1 μm |
13 mm | 13 mm | 13 mm | 47 mm |
Whatman 6809-7013, Whatman Article No. 28420420 (US reference) | Whatman 6809-7003, Whatman Article No. 28420419 (US reference) | Whatman 6809-7023, Whatman Article No. 28420421 (US reference) | Whatman 6809-5012, Whatman Article No. 28420412 (US reference) |
non-sterile | non-sterile | non-sterile | non-sterile |
pkg of 100 ea | pkg of 100 ea | pkg of 100 ea | pkg of 50 ea |
Analysis Note
BCR479
Other Notes
Nitrate (NO3)
Matrix Group: Beverages and Water
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service