Skip to Content
Merck
All Photos(1)

Key Documents

BCR308

9-Nitroanthracene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9NO2
CAS Number:
Molecular Weight:
223.23
Beilstein:
1877509
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

141-144 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1c2ccccc2cc3ccccc13

InChI

1S/C14H9NO2/c16-15(17)14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-9H

InChI key

LSIKFJXEYJIZNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

9-Nitroanthracene belongs to the class of nitrated-polycyclic aromatic hydrocarbons, found to be persistent in the environment. It is produced from direct sources such as diesel, gasoline exhaust and by the gas-phase reactions of PAHs with oxides of nitrogen.

Application

9-Nitroanthracene may be used as a certified reference material for the quantification of the analyte in the following:
  • Aerosol samples using direct infusion into an atmospheric pressure photoionization high-resolution mass spectrometer (APPI-HRMS).
  • Air and diesel particulate reference materials using pressurized-fluid extraction (PFE) with normal-phase liquid chromatography (LC) followed by gas chromatography coupled to mass spectrometry (GC-MS) in negative ion chemical ionization (NICI) mode.

Analysis Note

For more information please see:
BCR308

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct target and non-target analysis of urban aerosol sample extracts using atmospheric pressure photoionization high-resolution mass spectrometry
Giorio C, et al.
Chemosphere, 224, 786-795 (2019)
Andrea Alparone et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 89, 129-136 (2012-01-20)
Structure, IR and Raman spectra of 1-, 2- and 9-nitroanthracene isomers (1-NA, 2-NA and 9-NA) were calculated and analyzed through density functional theory computations using the B3LYP functional with the 6-311+G** basis set. Steric and π-conjugative effects determine the characteristic
9-Nitroanthracene.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 33, 179-185 (1984-04-01)
Mutagens in diesel exhaust particulate. Identification and direct activities of 6-nitrobenzo[a]pyrene, 9-nitroanthracene, 1-nitropyrene and 5h-phenanthro[4,5-bcd]pyran-5-one.
J N Pitts et al.
Mutation research, 103(3-6), 241-249 (1982-03-01)
Application of 9-nitroanthracene as a matrix for laser desorption/ionization analysis of fluorinated fullerenes.
Alexey V Streletskiy et al.
Rapid communications in mass spectrometry : RCM, 18(3), 360-362 (2004-02-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service