Skip to Content
Merck
All Photos(1)

Key Documents

BCR158

Benz[c]acridine

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11N
CAS Number:
Molecular Weight:
229.28
Beilstein:
154999
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2nc3c(ccc4ccccc34)cc2c1

InChI

1S/C17H11N/c1-3-7-15-12(5-1)9-10-14-11-13-6-2-4-8-16(13)18-17(14)15/h1-11H

InChI key

OAPPEBNXKAKQGS-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR158

Legal Information

BCR is a registered trademark of European Commission

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Tanaka et al.
Anticancer research, 16(5A), 2837-2841 (1996-09-01)
The mutagenicity of eleven methylbenz[c]acridines (methyl-B[c]ACRs) was examined by the direct Ames' Salmonella mutagenicity assay on the standard tester strains of TA97a, TA98, TA100 and TA102, each of which is specific to a certain type of mutation. The Benz[c]acridines (B[c]ACRs)
K Kamata et al.
Anticancer research, 9(3), 579-582 (1989-05-01)
Raman spectra of methyl substituted benz[c]acridines (BAc) in the solid state were recorded. Their assignments were proposed for the frequencies of Raman spectra observed. The purpose of this work was to elucidate the mechanisms of interaction between BAc and DNA
T Kurihara et al.
Anticancer research, 18(1A), 429-432 (1998-05-06)
On the calculated pi-spin density (rho pi) of benz[c]acridines [1-12], benz[c]acridines [2,8-12] with alternately value of pi-spin density (rho pi) on angular ring, large negative pi-spin density at C-1 and C-3 positions and large positive pi-spin density at C-2 and
T Kurihara et al.
Anticancer research, 16(5A), 2757-2765 (1996-09-01)
In order to clarify the effects of methyl substitution on the carcinogenic activity, each resonance energy (RE) of benz[c]acridines, benzo[a]phenothiazines, chrysene, and their methyl derivatives was calculated by Aihara's TRE theory. Some correlations seem to exist between the values of
H Sakagami et al.
Anticancer research, 15(6B), 2533-2540 (1995-11-01)
A series of phenothiazine, benzo[a]phenothiazine and benz[c]acridine derivatives were compared for their ability to induce nucleosome-sized DNA fragmentation (a biochemical hallmark of apoptosis), using agarose gel electrophoresis and a fluorescence activated cell sorter. Significant DNA fragmentation-inducing activity was detected in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service