Skip to Content
Merck
All Photos(1)

Key Documents

B9431

Sigma-Aldrich

Biotin-4-Fluorescein

≥95% purity (HPLC), powder

Synonym(s):

5-(6)-[2-(biotinoyl)aminoethylaminocarbonyl] fluorescein, B4F, Biotin (5-fluorescein) conjugate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H32N4O8S
Molecular Weight:
644.69
UNSPSC Code:
12171500
eCl@ss:
34058011
PubChem Substance ID:
NACRES:
NA.47

product name

Biotin-4-Fluorescein, ≥95.0% (HPLC)

Quality Level

100
200

Assay

≥95.0% (HPLC)

form

powder

mol wt

644.71

color

light orange to very dark orange
light yellow to very dark yellow

ε (extinction coefficient)

≥70 at 491-497 nm in phosphate buffer at 0.05 M

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OC(=O)c1cc(ccc1C2=C3C=CC(=O)C=C3Oc4ccc(O)cc24)C(=O)NCCNC(=O)CCCC[C@@H]5SC[C@@H]6NC(=O)N[C@H]56

Application

Biotin-4-Fluorescein has been used for the labeling of avidin to study the changes in protein binding capability after carbon negative-ion implantation into avidin. It has been used to study the binding affinity of streptavidin towards biotin.
Useful for quantitating biotin binding sites. This reagent binds to avidin more quickly than conventional fluorescein biotin.

Biochem/physiol Actions

Biotin-fluorescein is mainly used for the assay of biotin-interaction sites in avidin and streptavidin. Binding of biotin-4-fluorescein results in strong quenching, which is used for measuring the concentration of (strept)avidin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid estimation of avidin and streptavidin by fluorescence quenching or fluorescence polarization.
Kada G, et al.
Biochimica et Biophysica Acta, 1427, 44-48 (1999)
Sivaraj Sivaramakrishnan et al.
The Journal of cell biology, 187(1), 53-60 (2009-09-30)
Unconventional myosins interact with the dense cortical actin network during processes such as membrane trafficking, cell migration, and mechanotransduction. Our understanding of unconventional myosin function is derived largely from assays that examine the interaction of a single myosin with a
Cell-Adhesion Patterning by using Carbon Negative-Ion Implantation into Albumin and Avidin Layers on Polystyrene.
Sommani P, et al.
Transactions of the Materials Research Society of Japan, 36.3, 289-292 (2011)
Nathaniel G Hentz et al.
Journal of laboratory automation, 19(2), 153-162 (2013-09-14)
This study illustrates how optimization of both liquid-handling accuracy and precision is critical to assay performance. The study was designed to examine (1) liquid-handling performance and (2) the effect of liquid-handling variability on two types of in vitro biochemical assays

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service