Skip to Content
Merck
All Photos(3)

Key Documents

B5906

Sigma-Aldrich

Bacteriochlorophyll from Rhodopseudomonas sphaeroides

Powder

Synonym(s):

Bacteriochlorophyll a

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C55H74N4O6Mg
CAS Number:
Molecular Weight:
911.50
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Bacteriochlorophyll from Rhodopseudomonas sphaeroides,

form

powder

Quality Level

technique(s)

microbe id | staining: suitable

color

dark blue
light green to blue-green

solubility

ethanol: 10 mg/mL, light green

ε (extinction coefficient)

150-250 at 570-578 nm in diethyl ether

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

CC[C@@H]1[C@@H](C)c2cc3c(C(C)=O)c(C)c4cc5nc([C@@H](CCC(=O)OC\C=C(/C)CCC[C@H](C)CCC[C@H](C)CCCC(C)C)[C@@H]5C)c6[C@@H](C(=O)OC)C(=O)c7c(C)c(cc1n2)n([Mg]n34)c67

InChI

1S/C55H75N4O6.Mg/c1-13-39-34(7)41-29-46-48(38(11)60)36(9)43(57-46)27-42-35(8)40(52(58-42)50-51(55(63)64-12)54(62)49-37(10)44(59-53(49)50)28-45(39)56-41)23-24-47(61)65-26-25-33(6)22-16-21-32(5)20-15-19-31(4)18-14-17-30(2)3;/h25,27-32,34-35,39-40,51H,13-24,26H2,1-12H3,(H-,56,57,58,59,60,62);/q-1;+2/p-1/b33-25+;/t31-,32-,34-,35+,39-,40+,51-;/m1./s1

InChI key

DSJXIQQMORJERS-AGGZHOMASA-M

Looking for similar products? Visit Product Comparison Guide

General description

Bacteriochlorophylls are photosynthetic pigments, that are present in most of the bacteria.

Application

Bacteriochlorophyll from Rhodopseudomonas sphaeroides has been used as a standard to calibrate the high performance liquid chromatography (HPLC).

Biochem/physiol Actions

Bacteriochlorophyll/BchF or 3-vinyl-bacteriochlorophyll hydratase is an enzyme present in bacteria, which helps in producing bacteriochlorophyll a. Bacteriochlorophyll a is formed from chlorophyllide a.

Quality

Partially purified

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anticancer Drugs Acting via Radical Species: Radiotherapy and Photodynamic Therapy of Cancer
Medicinal Chemistry of Anticancer Drugs, 133-195 (2015)
Vertical gradient of nutrients in two dimictic lakes- influence of phototrophic sulfur bacteria on nutrient balance
Selig U, et al.
Aquatic sciences, 66(3), 247- 256 (2004)
Origin of bacteriochlorophyll a and the early diversification of photosynthesis
Cardona T
Testing, 11(3), e0151250-e0151250 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service