Skip to Content
Merck
All Photos(2)

Key Documents

A9199

Sigma-Aldrich

2-Amino-2-methyl-1-propanol

BioXtra, ≥95%

Synonym(s):

β-Aminoisobutyl alcohol, AMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(NH2)CH2OH
CAS Number:
Molecular Weight:
89.14
Beilstein:
505979
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

vapor density

3 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 25 °C)

product line

BioXtra

Assay

≥95%

form

liquid

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

refractive index

n20/D 1.4455 (lit.)

pH

11.0-12 (25 °C, 8.9 g/L)

useful pH range

9.0-10.5

pKa (25 °C)

9.7

bp

165 °C (lit.)

mp

24-28 °C (lit.)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

density

0.934 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

CC(C)(N)CO

InChI

1S/C4H11NO/c1-4(2,5)3-6/h6H,3,5H2,1-2H3

InChI key

CBTVGIZVANVGBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Amino-2-methyl-1-propanol has been used as an component in enzyme assay for screening the alkaline phosphatase activity in sarcoma osteogenic (SaOS-2) cells..

Biochem/physiol Actions

2-Amino-2-methyl-1-propanol is a substituted aliphatic alcohol and is used majorly as a pH balancer in cosmetic formulations. It has phototoxic effect as it can interact and penetrate above the sebum layer. However, it is not carcinogenic.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

179.8 °F - closed cup

Flash Point(C)

82.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The morphogenetically active polymer, inorganic polyphosphate complexed with GdCl3, as an inducer of hydroxyapatite formation in vitro
Wang X, et al.
Biochemical Pharmacology, 102(6), 97-106 (2016)
Final amended report on safety assessment on aminomethyl propanol and aminomethyl propanediol
Burnett CL, et al.
International Journal of Toxicology, 28(6), 141S-161S (2009)
Phototoxic assessment of a sunscreen formulation and its excipients: An in vivo and in vitro study
Hossy BH, et al.
Journal of Photochemistry and Photobiology. B, Biology, 173(6), 545-550 (2017)
Sontaya Limmatvapirat et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 67(3), 690-698 (2007-06-20)
The objective of this study was to improve the properties of shellac by composite salts formation. The shellac samples were prepared in various salt forms by dissolving them with 2-amino-2-methyl-1-propanol (AMP) and ammonium hydroxide (AMN) at various ratios of AMP:AMN.
Shia-Chung Chen et al.
Journal of environmental management, 92(4), 1083-1090 (2011-01-05)
In this study, the absorption of carbon dioxide using an absorbent composed of 2-amino-2-methyl-L-propanol (AMP) + monoethanolamine (MEA) + piperazine (PZ) in asymmetric and symmetric polytetrafluoroethylene (PTFE) membrane contactors was investigated. Experiments were conducted using various gas flow rates, liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service