Skip to Content
Merck
All Photos(5)

Key Documents

A2790

Sigma-Aldrich

Agarose

Wide range, for molecular biology

Synonym(s):

3,6-Anhydro-α-L-galacto-β-D-galactan

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41105317
PubChem Substance ID:
NACRES:
NA.25

biological source

algae (marine)

Quality Level

grade

for molecular biology

form

powder

technique(s)

electrophoresis: suitable

EEO

≤0.15

mp

≤65 °C

transition temp

gel point ≤35 °C

anion traces

sulfate (SO42-): ≤0.15%

foreign activity

DNase, RNase, none detected

SMILES string

O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O[C@@H]4O[C@@H]5[C@H]([C@@H](OC5)[C@@H]4O)O[C@@H]6O[C@@H]([C@@H]([C@@H]([C@H]6O)O)O)CO)O)O[C@H]2[C@H]3OC[C@@H]2O[C@H]([C@H]3O)O

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Wide range agarose is capable of separating DNA fragments from 50-1,000 bp on a single 3% agarose gel.

Analysis Note

The following is a list of properties associated with our agaroses:
Sulfate content - used as an indicator of purity, since sulfate is the major ionic group present.
Gel strength - the force that must be applied to a gel to cause it to fracture.
Gel point - the temperature at which an aqueous agarose solution forms a gel as it cools. Agarose solutions exhibit hysteresis in the liquid-to-gel transition - that is, their gel point is not the same as their melting temperature.
Electroendosmosis (EEO) - a movement of liquid through the gel. Anionic groups in an agarose gel are affixed to the matrix and cannot move, but dissociable counter cations can migrate toward the cathode in the matrix, giving rise to EEO. Since electrophoretic movement of biopolymers is usually toward the anode, EEO can disrupt separations because of internal convection.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Katherine R Long et al.
Neuron, 99(4), 702-719 (2018-08-07)
Neocortical expansion, thought to underlie the cognitive traits unique to humans, is accompanied by cortical folding. This folding starts around gestational week (GW) 20, but what causes it remains largely unknown. Extracellular matrix (ECM) has been previously implicated in neocortical
Sujith V W Weerasinghe et al.
Journal of medicinal chemistry, 51(18), 5542-5551 (2008-08-30)
Histone deacetylase 1 (HDAC1) has been linked to cell growth and cell cycle regulation, which makes it a widely recognized target for anticancer drugs. Whereas variations of the metal-binding and capping groups of HDAC inhibitors have been studied extensively, the
Michael J Lafferty et al.
The Journal of biological chemistry, 288(40), 28524-28534 (2013-08-21)
Elevated triglycerides are associated with an increased risk of cardiovascular disease, and lipoprotein lipase (LPL) is the rate-limiting enzyme for the hydrolysis of triglycerides from circulating lipoproteins. The N-terminal domain of angiopoietin-like protein 4 (ANGPTL4) inhibits LPL activity. ANGPTL4 was
Julia Dahlmann et al.
Biomaterials, 34(10), 2463-2471 (2013-01-22)
In most pluripotent stem cell differentiation protocols the formation of embryoid bodies (EBs) is an important step. Here we describe a rapid, straightforward soft lithography approach for the preparation of hydrophilic silicon masters from different templates and the subsequent production
Jia Liu et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(17), 6694-6699 (2013-04-10)
Seamless and minimally invasive integration of 3D electronic circuitry within host materials could enable the development of materials systems that are self-monitoring and allow for communication with external environments. Here, we report a general strategy for preparing ordered 3D interconnected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service