Skip to Content
Merck
All Photos(3)

Key Documents

A1662

Sigma-Aldrich

Bovine Serum Albumin solution

30%±2% in 0.85% sodium chloride, aseptically filled

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

bovine

Quality Level

sterility

aseptically filled

Assay

28-32% protein basis (biuret)

form

solution

purified by

heat shock fractionation

contains

0.1% sodium azide as preservative
10-15 mg/gram protein octanoic acid as stabilizer

origin

USA origin

concentration

28-32% (biuret)
30%±2% in 0.85% sodium chloride

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Bovine serum albumin (BSA), a non-glycosylated globular protein, is synthesized in the liver without the use of prosthetic groups or other additives, making it one of the few plasma proteins without carbohydrate groups. BSA reveals distinct binding sites with varying specificities. Site-I and site-II are the most important ones, located in the hydrophobic cavities of subdomains IIA and IIIA, respectively.

Application

Bovine Serum Albumin solution has been used:
  • in the incubation of 108 colony-forming units S. aureus for quantitative suspension assay to evaluate the antibacterial efficacy
  • as a component in bacterial broth culture as organic matter loading for control experiments to study the microbicidal impact of the sesamoid bone against Staphylococcus aureus
  • as organic matter loading in Ham′s F12 culture medium for quantitative suspension tests to observe the effect of antimicrobial action of the antiseptics

Biochem/physiol Actions

Bovine serum albumin (BSA) functions as a transport carrier for drugs. It is primarily used as a test model for studying albumin interactions with strong pharmaceutical agents. The ability of BSA to interact with many aromatic and hydrophobic molecules in the bloodstream makes it useful. In cell culture media, bovine serum albumin is frequently used as a protein supplement. The fluorescent property of BSA aids in emitting intensely upon excitation.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The NMR tube bioreactor
Chatzikonstantinou AV, et al.
Test, 633, 71-101 (2020)
Bovine serum albumin
2018 IEEE International Interconnect Technology Conference (IITC), 1045-1045 (2016)
N Kapalschinski et al.
Burns : journal of the International Society for Burn Injuries, 39(6), 1221-1225 (2013-05-15)
Wound infection is one of the major complications in acute and chronic wound healing. Antiseptic solutions and wound irrigating agents are routinely used for therapy and prevention in healthcare today. Even if wound exudate contains total protein concentrations up to
Gerald Müller et al.
Chemico-biological interactions, 145(3), 331-336 (2003-05-07)
Anatomically intact articular cartilage in form of sesamoid bones from metacarpophalangeal joints of 2-year-old cows was tested for its influence on the microbicidal effect of the iodophore Betaisodona, the bispyridinamine Octenisept, and the biguanide Lavasept. Comparisons were carried out in
The NMR tube bioreactor
Chatzikonstantinou AV, et al.
Methods in Enzymology, 633, 71-101 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service