Skip to Content
Merck
All Photos(4)

Key Documents

A1028

Sigma-Aldrich

8-Anilino-1-naphthalenesulfonic acid

≥97% purity (HPLC), powder

Synonym(s):

N-Phenyl peri acid, ANS, ANSA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHC10H6SO3H
CAS Number:
Molecular Weight:
299.34
Beilstein:
1843887
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

8-Anilino-1-naphthalenesulfonic acid,

Assay

≥97% (HPLC)

Quality Level

form

powder

composition

, <0.5% (solvent)

color

black
gray to dark green

mp

215-217 °C (dec.) (lit.)

solubility

DMF: 50 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OS(=O)(=O)c1cccc2cccc(Nc3ccccc3)c12

InChI

1S/C16H13NO3S/c18-21(19,20)15-11-5-7-12-6-4-10-14(16(12)15)17-13-8-2-1-3-9-13/h1-11,17H,(H,18,19,20)

InChI key

FWEOQOXTVHGIFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

8-Anilino-1-naphthalenesulfonic acid (ANS) has been used to study the thermal aggregation of BSA (bovine serum albumin).

Biochem/physiol Actions

8-Anilino-1-naphthalenesulfonic acid (ANS) is an important dye which is used to study the intermediate states in the folding/unfolding pathways of proteins. It is also useful to check the conformational changes involved with the aggregation of proteins. ANS is nonfluorescent in the aqueous solution, however, it fluoresces strongly upon binding to BSA (bovine serum albumin) and denatured proteins. It is used for the detection of albumin in the serum. ANS mainly interacts with the solvent exposed nonpolar (hydrophobic) sites.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal aggregation of glycated bovine serum albumin.
Rondeau P, et al.
Biochimica et Biophysica Acta, 1804, 789-789 (2010)
Uversky VN and Permiakov EA
Methods in Protein Structure and Stability Analysis (2007)
Lauren E Schrock et al.
Movement disorders : official journal of the Movement Disorder Society, 30(4), 448-471 (2014-12-06)
Deep brain stimulation (DBS) may improve disabling tics in severely affected medication and behaviorally resistant Tourette syndrome (TS). Here we review all reported cases of TS DBS and provide updated recommendations for selection, assessment, and management of potential TS DBS
Thermal aggregation of bovine serum albumin at different pH: comparison with human serum albumin.
Vetri V, et al.
European Biophysics Journal, 36, 717-717 (2007)
Michelle J Chandley et al.
Molecular autism, 6, 28-28 (2015-05-23)
The anterior cingulate cortex (ACC) is a brain area involved in modulating behavior associated with social interaction, disruption of which is a core feature of autism spectrum disorder (ASD). Functional brain imaging studies demonstrate abnormalities of the ACC in ASD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service