Skip to Content
Merck
All Photos(1)

Key Documents

95148

Supelco

L-Argininosuccinic acid lithium salt

analytical standard

Synonym(s):

N-{{[(4S)-4-Amino-4-carboxybutyl]amino}iminomethyl}-L-aspartic acid lithium salt, Lithium L-argininosuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18N4O6 · xLi+
Molecular Weight:
290.27 (free acid basis)
Beilstein:
6933810
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Assay

≥90.0% (HPLC)

form

solid

optical activity

[α]/D 15±2°, c = 1 in H2O

shelf life

limited shelf life, expiry date on the label

impurities

≤20% water

application(s)

clinical testing

storage temp.

2-8°C

SMILES string

OC(C[C@@H](C(O)=O)NC(NCCC[C@H](N)C(O)=O)=N)=O.[Li+]

InChI

1S/C10H18N4O6.Li/c11-5(8(17)18)2-1-3-13-10(12)14-6(9(19)20)4-7(15)16;/h5-6H,1-4,11H2,(H,15,16)(H,17,18)(H,19,20)(H3,12,13,14);/q;+1/t5-,6-;/m0./s1

InChI key

MGJRREWOOITZBB-GEMLJDPKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Arginosuccinic acid is a basic amino acid, which is synthesized by some cells from citrulline, aspartic acid and used as a precursor for arginine in the urea cycle or Citrulline-NO cycle. The reaction is catalyzed by the enzyme argininosuccinate synthetase. Argininosuccinic acid is a precursor to fumarate in the citric acid cycle via argininosuccinate lyase. Defects in the arginosuccinate lyase enzyme can lead to arginosuccinate lyase deficiency. Argininosuccinate (ASA) lyase deficiency results in defective cleavage of ASA. This leads to an accumulation of ASA in cells and an excessive excretion of ASA in urine (arginosuccinic aciduria). In virtually all respects, this disorder shares the characteristics of other urea cycle defects. The most important characteristic of ASA lyase deficiency is its propensity to cause hyperammonemia in affected individuals. ASA in affected individuals is excreted by the kidney at a rate practically equivalent to the glomerular filtration rate (GFR). Whether ASA itself causes a degree of toxicity due to hepatocellular accumulation is unknown; such an effect could help explain hyperammonemia development in affected individuals. Regardless, the name of the disease is derived from the rapid clearance of ASA in urine, although elevated levels of ASA can be found in plasma. ASA lyase deficiency is associated with high mortality and morbidity rates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service