Skip to Content
Merck
All Photos(1)

Key Documents

94685

Supelco

Vanadate-molybdate reagent

p.a.

Synonym(s):

VM reagent

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12164500
NACRES:
NA.84

grade

p.a.

Quality Level

form

liquid

specific analyte(s)

phosphate

technique(s)

photometry: suitable

impurities

sulfuric acid

density

1.15 g/mL at 20 °C

application(s)

general analytical

General description

Vanadate-molybdate reagent can be prepared by dissolving ammonium molybdate and ammonium vanadate in distilled water along with concentrated nitric acid.

Application

Vanadate-molybdate reagent has been used to determine phosphorous using atomic absorption spectrometer, by vanadium phosphomolybdate method from cashew apple juice samples.

Other Notes

Reagent, suitable for eliminating interfering effects by phosphate and silicate in the AAS determination of Ca and Mg

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I. Sarudi et al.
Fresenius Journal of Analytical Chemistry, 321, 493-493 (1985)
I. Sarudi
Fresenius Journal of Analytical Chemistry, 303, 197-197 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service