Skip to Content
Merck
All Photos(1)

Key Documents

94226

Millipore

Raffinose Disks

suitable for microbiology, Sterile filter paper discs impregnated with raffinose

Synonym(s):

Carbohydrate discs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

Quality Level

sterility

sterile

product line

BioChemika

form

disc

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 10 × 25 discs

technique(s)

microbe id | utilization test: suitable

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Enterobacter spp.
Klebsiella spp.
bacteria

General description

The raffinose fermentation test is based on the fermentation ability of raffinose which results in the production of acid as end products, the pH of the liquid basal medium will drop, and an indicator changes the color to red. Some organisms produce additional CO2 gas which can be made visible with a Durham’s tube. Raffinose can be used to differentiate Enterobacteriaceae, Bifidobacteria and Lactobacilli, Enterococcus faecium and Streptococcus pneumoniae can utilize that sugar.

Application

Raffinose Discs are recommended to detect raffinose fermenting bacteria in food and environmental samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service