Skip to Content
Merck
All Photos(2)

Key Documents

93465

Supelco

Ethylhexyl triazone

analytical standard

Synonym(s):

2,4,6-Trianilino(p-carbo-2-ethylhexyl-1-oxy)-1,3,5-triazine, Tris(2-ethylhexyl) 4,4′,4″-(1,3,5-triazine-2,4,6-triyltriimino)trisbenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H66N6O6
CAS Number:
Molecular Weight:
823.07
Beilstein:
11001015
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5% water (calc. from elemental analysis)

mp

128-132 °C

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCC(CC)COC(=O)c1ccc(Nc2nc(Nc3ccc(cc3)C(=O)OCC(CC)CCCC)nc(Nc4ccc(cc4)C(=O)OCC(CC)CCCC)n2)cc1

InChI

1S/C48H66N6O6/c1-7-13-16-34(10-4)31-58-43(55)37-19-25-40(26-20-37)49-46-52-47(50-41-27-21-38(22-28-41)44(56)59-32-35(11-5)17-14-8-2)54-48(53-46)51-42-29-23-39(24-30-42)45(57)60-33-36(12-6)18-15-9-3/h19-30,34-36H,7-18,31-33H2,1-6H3,(H3,49,50,51,52,53,54)

InChI key

JGUMTYWKIBJSTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylhexyl triazone has a high absorption coefficient and photostability, which makes it an important ingredient in cosmetic formulations, where high SPF value is required.
Ethylhexyl triazone is a ultraviolet-B filter, which is used as a sunscreen agent in skin care products.

Application

Ethylhexyl triazone may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Cosmetic samples using ultra-high-performance supercritical fluid chromatography (UHPSFC).
  • Topical skincare products using normal-phase high-performance thin layer chromatography (HPTLC) and densitometry.
  • Environmental samples using liquid chromatography-tandem mass spectrometry (LC-MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

584.6 °F

Flash Point(C)

307 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoallergic contact dermatitis from the sunscreen octyl triazone.
S Sommer et al.
Contact dermatitis, 46(5), 304-305 (2002-06-27)
Cécile Plagellat et al.
Chemosphere, 62(6), 915-925 (2005-07-06)
Many substances related to human activities end up in wastewater and accumulate in sewage sludge. The present study focuses on the analysis of widely used UV filters 3-(4-methylbenzylidene) camphor (4-MBC), octyl-methoxycinnamate (OMC), octocrylene (OC) and octyl-triazone (OT) in sewage sludge
Novel emerging sunscreen technologies
Tuchinda C, et al.
Dermatologic Clinics, 24, 105-117 (2006)
Solution thermodynamics of ethylhexyl triazone in some ethanol + ethyl acetate mixtures
Rodriguez.JS, et al.
Journal of Solution Chemistry, 39, 1122-1133 (2010)
Anna W Sobanska et al.
TheScientificWorldJournal, 2012, 807516-807516 (2012-05-26)
Ethylhexyl triazone (ET) was separated from other sunscreens such as avobenzone, octocrylene, octyl methoxycinnamate, and diethylamino hydroxybenzoyl hexyl benzoate and from parabens by normal-phase HPTLC on silica gel 60 as stationary phase. Two mobile phases were particularly effective: (A) cyclohexane-diethyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service