Skip to Content
Merck
All Photos(1)

Key Documents

91332

Sigma-Aldrich

2,4,5-Trichlorophenol

purum, ≥95.0% (GC)

Synonym(s):

TCP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2OH
CAS Number:
Molecular Weight:
197.45
Beilstein:
607569
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥95.0% (GC)

form

powder

bp

248 °C/740 mmHg (lit.)

mp

64-67 °C
67-69 °C (lit.)

functional group

chloro

SMILES string

Oc1cc(Cl)c(Cl)cc1Cl

InChI

1S/C6H3Cl3O/c7-3-1-5(9)6(10)2-4(3)8/h1-2,10H

InChI key

LHJGJYXLEPZJPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

271.4 °F - closed cup

Flash Point(C)

133.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Bello et al.
Environmental pollution (Barking, Essex : 1987), 156(3), 611-617 (2008-07-29)
Soil response to contamination with 2,4,5-triclorophenol was studied to test the validity of the concept of Generic Reference Levels (GRL), the main criterion used to define soil contamination. Soil samples were artificially contaminated with doses between 0 and 5000 mg
Qiao Sun et al.
Environmental science & technology, 41(16), 5708-5715 (2007-09-19)
Density functional theory (DFT) calculations have been carried out to explore the potential energy surface (PES) associated with the gas-phase reaction between 2,4,5-trichlorophenol and CuO. A gas-phase model was constructed to account, from a theoretical perspective, for the most important
Xiaohui Xu et al.
Occupational and environmental medicine, 68(8), 557-561 (2011-05-05)
Trichlorophenols (TCPs) are organochlorine compounds which are ubiquitous in the environment and well known for their carcinogenic effects. However, little is known about their neurotoxicity in humans. Our goal was to examine the association between body burden of TCPs (ie
Mohamed Abdel Salam et al.
Analytical and bioanalytical chemistry, 390(8), 2159-2170 (2008-03-13)
This paper describes a novel application of pristine and chemically modified multiwalled carbon nanotubes (MWCNTs) as the packing materials for the determination of different polyhalogenated organic pollutants, pentachlorophenol, 2,4,5-trichlorophenol, 3,3',4,4'-tetrachlorobiphenyl and 2,2',5,5'-tetrabromobiphenyl, from aqueous solution based on solid phase extraction.
James J Collins et al.
Journal of exposure science & environmental epidemiology, 17(6), 541-548 (2007-04-12)
This study examines serum levels of 2,3,7,8-substituted chlorinated dioxins and furans, and PCBs for 375 Michigan workers with potential chlorophenol exposure, 37 Worker Referents, and 71 Community Referents. The chlorophenol workers were last exposed to trichlorophenol and/or pentachlorophenol 26-62 years

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service