Skip to Content
Merck
All Photos(1)

Key Documents

90383

Supelco

Chlorotriethylsilane

for GC derivatization, LiChropur, ≥97.0% (GC)

Synonym(s):

TESCl, Triethylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3SiCl
CAS Number:
Molecular Weight:
150.72
Beilstein:
1732860
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥97.0% (GC)

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.43 (lit.)

bp

142-144 °C (lit.)

density

0.898 g/mL at 25 °C (lit.)

SMILES string

CC[Si](Cl)(CC)CC

InChI

1S/C6H15ClSi/c1-4-8(7,5-2)6-3/h4-6H2,1-3H3

InChI key

DCFKHNIGBAHNSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorotriethylsilane may be used as a silylating reagent for the preparation of trisilyl ethers from alcohols in the presence of a base such as imidazole, pyridine or 4-(dimethylamino)pyridine. It is found to be more reactive when used with pyridine as a solvent.

Application

Chlorotriethylsilane may be used as a derivatizing reagent for the determination of fluoride, in toothpaste samples using headspace solid-phase microextraction and gas chromatography–flame ionization detection.

Other Notes

Silylating agent with some advantage over the trimethylsilylating agents for synthetic and analytical applications

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

86.0 °F

Flash Point(C)

30 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 4 (2014)
Determination of fluoride in toothpaste using headspace solid-phase microextraction and gas chromatography?flame ionization detection
Wejnerowska G, et al.
Journal of Chromatography A, 1150(1-2), 173-177 (2007)
W.C. Still et al.
Journal of the American Chemical Society, 99, 948-948 (1977)
J. Heberle et al.
Silylating Agents, 2nd ed. (1995)
C.F. Poole et al.
Journal of Chromatographic Science, 17, 115-115 (1979)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service