Skip to Content
Merck
All Photos(1)

Key Documents

90240

Sigma-Aldrich

Glyceryl triacetate

≥99.0% (GC)

Synonym(s):

1,2,3-Triacetoxypropane, 1,2,3-Triacetylglycerol, Glycerol triacetate, Triacetin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COOCH2)2CHOCOCH3
CAS Number:
Molecular Weight:
218.20
Beilstein:
1792353
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99.0% (GC)

form

liquid

refractive index

n20/D 1.431

bp

258-260 °C (lit.)

density

1.158 g/mL at 20 °C

lipid type

saturated FAs

SMILES string

CC(=O)OCC(COC(C)=O)OC(C)=O

InChI

1S/C9H14O6/c1-6(10)13-4-9(15-8(3)12)5-14-7(2)11/h9H,4-5H2,1-3H3

InChI key

URAYPUMNDPQOKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Epigenetic priming targets tumor heterogeneity to shift transcriptomic phenotype of pancreatic ductal adenocarcinoma towards a Vitamin D susceptible state.: This groundbreaking study utilizes glyceryl triacetate as part of an epigenetic priming strategy to sensitize pancreatic ductal adenocarcinoma to Vitamin D, marking a significant advancement in cancer treatment modalities (He et al., 2024).

  • Glyceryl triacetate promotes blood-brain barrier recovery after ischemic stroke through lipogenesis-mediated IL-33 in mice.: Research demonstrates glyceryl triacetate′s effectiveness in enhancing blood-brain barrier repair post-ischemic stroke via lipogenesis pathways, offering insights into novel stroke recovery treatments (Wei et al., 2023).

  • Development of A Nanostructured Lipid Carrier-Based Drug Delivery Strategy for Apigenin: Experimental Design Based on CCD-RSM and Evaluation against NSCLC In Vitro.: This study details the formulation of a nanostructured lipid carrier incorporating glyceryl triacetate, optimized for delivering apigenin to combat non-small cell lung cancer, emphasizing the potential of glyceryl triacetate in targeted therapy applications (Wang et al., 2023).

  • Are New Phthalate Ester Substitutes Safer than Traditional DBP and DiBP Comparative Endocrine-Disrupting Analyses on Zebrafish Using In Vivo, Transcriptome, and In Silico Approaches.: Investigating glyceryl triacetate as a safer alternative to traditional phthalate esters, this study explores its lower endocrine-disrupting effects, contributing to safer industrial and pharmaceutical applications (Tan et al., 2023).

  • Glyceryl triacetate feeding in mice increases plasma acetate levels but has no anticonvulsant effects in acute electrical seizure models.: This research evaluates the biochemical impact of glyceryl triacetate on plasma acetate levels, providing valuable data on its metabolic effects despite a lack of anticonvulsant properties in specific seizure models (Xu et al., 2022).


Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Liu et al.
Journal of pharmaceutical sciences, 101(5), 1783-1793 (2012-02-10)
Although injectable depot-forming solutions have been commercialized, the factors that influence the overall release kinetics from such systems are still not fully understood. In this work, we address the effect of cosolvent on the issue of excessive burst release of
Huayin Pu et al.
Journal of agricultural and food chemistry, 59(10), 5738-5745 (2011-04-26)
An oral colon-targeting controlled release system based on resistant starch acetate (RSA) as a film-coating material was developed. The RSA was successfully synthesized, and its digestion resistibility could be improved by increasing the degree of substitution (DS), which was favorable
Seung Jun Choi et al.
Journal of agricultural and food chemistry, 57(23), 11349-11353 (2009-11-07)
Citral is widely used in the beverage, food, and fragrance industries for its characteristic flavor profile. However, it chemically degrades over time in aqueous solutions due to an acid-catalyzed reaction, which leads to loss of desirable flavor notes and formation
Miguel Castañeda-Zárate et al.
Current biology : CB, 31(1), 238-246 (2020-11-07)
During the evolutionary history of flowering plants, transitions between pollinator groups (pollinator shifts) have been frequent,1 and contributed to the spectacular radiation of angiosperms.2 Although the evolution of floral traits during pollinator shifts has been studied in real time under
Orit Amsalem et al.
Colloids and surfaces. B, Biointerfaces, 81(2), 422-429 (2010-08-17)
Complex pseudo-ternary phase diagrams based on sucrose monolaurate (SE), propylene glycol (PG), and phosphatidylcholine (PC) as the "surfactant phase"; triacetin (TA) and decaglycerol ester (10G1CC) as the "oil phase"; and water as the aqueous phase were constructed, into which we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service