Skip to Content
Merck
All Photos(1)

Key Documents

902000

Sigma-Aldrich

UltraNitroCat

≥95%

Synonym(s):

(1-(2,6-Diethylphenyl)-3,5,5-trimethyl-3-phenylpyrrolidin-2-ylidene)dichloro(2-isopropoxy-5-nitrobenzylidene)ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40Cl2N2O3Ru
CAS Number:
Molecular Weight:
684.66
UNSPSC Code:
12352101

Assay

≥95%

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

availability

available only in USA

mp

>300 °C

storage temp.

2-8°C

Application

UltraNitroCat is efficient in macrocyclization. Reactions can be run at 30 to 85 °C. The following reaction solvents are compatible: toluene, ethyl acetate, dimethylcarbonate, and CPME (cyclopentyl methyl ether).

Legal Information

Sold in collaboration with Apeiron Synthesis, Inc. Research quantities are 100 g / customer / year. For quantities that exceed the research limit, please contact the supplier at www.apeiron-synthesis.com.
WO 2017/055945

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service