Skip to Content
Merck
All Photos(1)

Key Documents

900632

Sigma-Aldrich

Triethylamine

ZerO2®, ≥99%

Synonym(s):

Degassed and low oxygen triethylamine, Triethylamine ZerO2®, N,N-Diethylethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
Beilstein:
605283
EC Number:
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21
grade:
ZerO2®
bp:
88.8 °C (lit.)
vapor pressure:
51.75 mmHg ( 20 °C)

grade

ZerO2®

Quality Level

vapor density

3.5 (vs air)

vapor pressure

51.75 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

593 °F

expl. lim.

8 %

impurities

≤0.5% water
water

refractive index

n20/D 1.401 (lit.)

pH

12.7 (15 °C, 100 g/L)

bp

88.8 °C (lit.)

mp

−115 °C (lit.)

density

0.726 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CC

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ZerO2 products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Legal Information

ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Hobeika et al.
BMC pharmacology & toxicology, 21(1), 15-15 (2020-02-24)
In developing countries, brand-generic substitution is not based on validated scientific evidence that confirm the therapeutic equivalence of the generic to the originator. Rather, decisions are made based on the availability of generic medications. Substitution by inappropriate preparations applies to

Articles

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service