Skip to Content
Merck
All Photos(2)

Key Documents

89505

Supelco

mono-Benzyl phthalate

analytical standard

Synonym(s):

1,2-Benzenedicarboxylic acid 1-(phenylmethyl) ester, Phthalic acid monobenzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O4
CAS Number:
Molecular Weight:
256.25
Beilstein:
1884373
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

mp

103-108 °C

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

OC(C1=C(C(OCC2=CC=CC=C2)=O)C=CC=C1)=O

InChI

1S/C15H12O4/c16-14(17)12-8-4-5-9-13(12)15(18)19-10-11-6-2-1-3-7-11/h1-9H,10H2,(H,16,17)

InChI key

XIKIUQUXDNHBFR-UHFFFAOYSA-N

General description

mono-Benzyl phthalate (MBzP) is a hydroxylated phthalate metabolite, identified as a major intermediate in the titanium dioxide (TiO2)/UV degradation of n-butyl benzyl phthalate (BBP). It is an endocrine disrupting compound and a priority pollutant found in diverse environmental samples. It finds applications as an industrial solvent and additive in a variety of products including adhesives, vinyl flooring products, car-care products, personal care products, etc. MBzP serves as an indicator or marker to determine the exposure to benzylbutyl phthalate (BBP).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
mono-Benzyl phthalate may be used as an analytical reference standard for the quantification of the analyte in:
  • Biological samples using high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS).
  • River water samples using gas chromatography coupled to mass spectrometry (GC-MS).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelly K Ferguson et al.
Environmental research, 111(5), 718-726 (2011-02-26)
Phthalate esters are a class of compounds utilized extensively in widely-distributed consumer goods, and have been associated with various adverse health outcomes in previous epidemiologic research. Some of these health outcomes may be the result of phthalate-induced increases in oxidative
Urinary phthalate metabolites and male reproductive function parameters in Chongqing general population, China
Han X, et al.
International Journal of Hygiene and Environmental Health, 217(2-3), 271-278 (2014)
Lizbeth López-Carrillo et al.
Environmental health perspectives, 118(4), 539-544 (2010-04-07)
Phthalates, ubiquitous environmental pollutants that may disturb the endocrine system, are used primarily as plasticizers of polyvinyl chloride and as additives in consumer and personal care products. In this study, we examined the association between urinary concentrations of nine phthalate
Monitoring of phthalic acid monoesters in river water by solid-phase extraction and GC-MS determination
Suzuki T, et al.
Environmental Science & Technology, 35(18), 3757-3763 (2001)
Sharon Daniel et al.
Environment international, 143, 105894-105894 (2020-07-18)
Emerging evidence suggests that phthalate exposure may be associated with behavior problems in children and that these associations may be sex specific. In a follow up study of 411 inner-city minority mothers and their children, mono-n-butyl phthalate (MnBP), monobenzyl phthalate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service