86626
Tetraethylammonium hydrogen sulfate
suitable for ion pair chromatography, LiChropur™, ≥99.0%
About This Item
description
cationic
Quality Level
Assay
≥99.0% (T)
≥99.0%
form
crystals
quality
LiChropur™
technique(s)
ion pair chromatography: suitable
mp
243-245 °C
λ
10 % in H2O
UV absorption
λ: 210 nm Amax: 0.05
λ: 220 nm Amax: 0.04
λ: 230 nm Amax: 0.03
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02
suitability
corresponds to standard for RP gradient test
corresponds to standard for filter test
SMILES string
OS([O-])(=O)=O.CC[N+](CC)(CC)CC
InChI
1S/C8H20N.H2O4S/c1-5-9(6-2,7-3)8-4;1-5(2,3)4/h5-8H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1
InChI key
CREVBWLEPKAZBH-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service