Skip to Content
Merck
All Photos(1)

Key Documents

85067

Sigma-Aldrich

Sesame oil from Sesamum indicum

tested according to Ph. Eur.

Synonym(s):

Sesami oleum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.21

biological source

Sesamum indicum (sesame) seeds

Quality Level

Agency

USP/NF
tested according to Ph. Eur.

form

liquid

refractive index

n20/D 1.474

functional group

oleic acid

lipid type

oils

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application


  • Efficacy and classification of Sesamum indicum linn seeds with Rosa damascena mill oil in uncomplicated pelvic inflammatory disease using machine learning.: This study applies machine learning to evaluate the efficacy of sesame seeds combined with rose oil in treating pelvic inflammatory disease, offering a novel therapeutic approach (Sumbul et al., 2024).

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

491.0 °F

Flash Point(C)

255 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Douglas D Reh et al.
The Laryngoscope, 123(4), 820-822 (2013-02-13)
Topical Sesame/Rose geranium oil compound is an effective therapy for hereditary hemorrhagic telangiectasia (HHT) associated epistaxis. Cohort Study. Twenty patients with HHT confirmed by the Curaçao criteria were treated with a sesame/rose geranium oil topical compound between January 2010 and
Srinivasan Periasamy et al.
JPEN. Journal of parenteral and enteral nutrition, 37(1), 129-133 (2012-05-01)
The chemotherapeutic drug oxaliplatin causes sinusoidal obstruction syndrome (SOS), which is analogous to that of monocrotaline-induced SOS. Sesame oil is a nutrient-rich antioxidant in alternative medicine. It contains phenol, sesamin, sesamol, and sesamolin, all of which contribute to its antioxidant
Vinay Dubey et al.
Applied biochemistry and biotechnology, 168(5), 1265-1276 (2012-09-14)
Probiotics with ability to produce conjugated linoleic acid (CLA) is considered as an additive health benefit property for its known role in colon cancer mitigation. The conversion involves the biohydrogenation of the unsaturated fatty acid into conjugated form. Probiotic strain
Kalliopi Karatzi et al.
Journal of clinical hypertension (Greenwich, Conn.), 14(9), 630-636 (2012-09-06)
The possible effects of sesame oil on hemodynamics are unknown. The aim of the study was to investigate the acute and long-term effects of sesame oil on hemodynamic responses in hypertensive men. The authors enrolled 30 hypertensive men in a
Kalliopi Karatzi et al.
European journal of preventive cardiology, 20(2), 202-208 (2012-02-22)
The aim of the study was to investigate the effects of sesame oil on endothelial function and to detect the underlying mechanisms, both in the postprandial state and after long-term consumption. We enrolled 30 hypertensive men in a two-phase study.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service