Skip to Content
Merck
All Photos(4)

Key Documents

84230

Sigma-Aldrich

Salicylamide

puriss., ≥99.0% (T)

Synonym(s):

2-Hydroxybenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CONH2
CAS Number:
Molecular Weight:
137.14
Beilstein:
742439
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

puriss.

Quality Level

Assay

≥99.0% (T)

form

solid

mp

139-141 °C
140-144 °C (lit.)

solubility

methanol: 0.1 g/mL, clear

functional group

amide

SMILES string

NC(=O)c1ccccc1O

InChI

1S/C7H7NO2/c8-7(10)5-3-1-2-4-6(5)9/h1-4,9H,(H2,8,10)

InChI key

SKZKKFZAGNVIMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Dolezal et al.
European journal of medicinal chemistry, 44(2), 869-876 (2008-06-13)
Quantitative relationships between the molecular structure and the biological activity of 49 isosteric salicylamide derivatives as potential antituberculotics with a new mechanism of action against three Mycobacterial strains were investigated. The molecular structures were represented by quantum chemical B3LYP/6-31G( *)
J A Murillo Pulgarín et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 909-914 (2011-05-13)
A new methodology for the simultaneous determination of salicylic acid and salicylamide in biological fluids is proposed. The strong overlapping of the fluorescence spectra of both analytes makes impossible the conventional fluorimetric determination. For that reason, the use of fluorescence
S Zhang et al.
Transplantation proceedings, 38(5), 1247-1252 (2006-06-27)
Cardiac ischemia/reperfusion (I/R) injury, a necessary consequence of transplantation, is probably related to the formation of reactive oxygen species (ROS). The ROS burst within the first moments of reperfusion is associated with injury, continuously generate O2- at about 3% to
Stephanie S Barnes et al.
Dalton transactions (Cambridge, England : 2003), 40(17), 4707-4714 (2011-03-25)
Addition of boranes to N-aryl-salicylaldimines takes place initially at the reactive phenolic O-H bond to give an activated boron-containing imine and dihydrogen. In some cases a subsequent intramolecular hydrogenation step is observed and the C=N imine bond is reduced to
Silvia Wein et al.
Xenobiotica; the fate of foreign compounds in biological systems, 42(5), 477-482 (2011-12-23)
We investigated acute effects and effects after chronic intake of the orally administered flavonol quercetin on pharmacokinetics of salicylamide metabolites (SAM) after oral administration of salicylamide in pigs. Salicylamide (8 mg/kg body weight) was orally administered to seven pigs either

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service