Skip to Content
Merck
All Photos(1)

Key Documents

82962

Millipore

Peptone from meat, enzymatic digest

for biotechnological purposes

Synonym(s):

Peptone from bovine meat

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
eCl@ss:
42040102
NACRES:
NA.74

biological source

Porcine pancreas
bovine heart (
)

Quality Level

form

powder

composition

amino-nitrogen, ≥2.7%
total nitrogen (N), ≥14%

packaging

pkg of (50 g

1 kg

5 kg)

ign. residue

≤6%

loss

≤6% loss on drying

pH

5.5±0.5 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear, faintly to light yellow

application(s)

food and beverages
microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Peptone from meat, enzymatic digest is a richsource of peptides and free amino acids. Meat peptone provides nitrogen,vitamins, amino acids, and carbon in microbiological culture media. High sulfurcompound content in meat peptone enables the detection of bacteria (Clostridiaspp., Salmonella spp.) that produce hydrogen sulfide. Meat peptone issuitable for the cultivation of yeast and molds, enterobacteria, andstaphylococci. It also favors the growth of streptococci and the production ofcharacteristic hemolysis in blood agar formulations.

Application

Peptone (from meat) is used as a nutritive substrate in fermentation and culture media. It is suitable for analytical microbiology and industrial fermentation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Strategies for high titre plasmid DNA production
in Escherichia coli DH5a
Kevin O?Mahony
Process. Biochem. (2007)
Hongmin Cai et al.
Protein expression and purification, 164, 105463-105463 (2019-08-06)
Recombinant expression of human membrane proteins in large quantities remains a major challenge. Expression host is an important variable to screen for high-level production of membrane proteins. Using the green fluorescent protein (GFP) as a reporter, we screened the expression
Proposal for a better integration of bacterial lysis into the
production of plasmid DNA at large scale
Kevin O?Mahony
Journal of Biotechnology (2005)
Hebang Yao et al.
Journal of molecular biology, 432(18), 5162-5183 (2020-02-28)
Membrane proteins are generally challenging to work with because of their notorious instability. Protein engineering has been used increasingly to thermostabilize labile membrane proteins such as G-protein-coupled receptors for structural and functional studies in recent years. Two major strategies exist.

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service