Skip to Content
Merck
All Photos(1)

Key Documents

81619

Sigma-Aldrich

Ectoine

≥95.0% (HPLC)

Synonym(s):

(S)-2-Methyl-1,4,5,6-tetrahydropyrimidine-4-carboxylic acid, Thp(B)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N2O2
CAS Number:
Molecular Weight:
142.16
Beilstein:
7288977
EC Number:
MDL number:
UNSPSC Code:
24112111
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥95.0% (HPLC)

form

powder

mol wt

142.16 g/mol

storage condition

dry at room temperature

color

white

solubility

methanol: soluble 10 mg/mL, clear, colorless to almost colorless

SMILES string

CC1=N[C@@H](CCN1)C(O)=O

InChI

1S/C6H10N2O2/c1-4-7-3-2-5(8-4)6(9)10/h5H,2-3H2,1H3,(H,7,8)(H,9,10)/t5-/m0/s1

InChI key

WQXNXVUDBPYKBA-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ectoine, also known as 4,5,6-tetrahydro-2-methyl-4-pyrimidinecarboxylic acid, is a zwitterionic amino acid derivative that is a compatible osmoprotectant solute produced in certain aerobic, chemoheterotrophic, and halophilic bacteria to protect against osmotic stress, radiation, and other unfavorable environmental conditions.

Application

The practical applications of ectoine include enzyme stabilization, anti-inflammatory treatment, neurodegenerative disease prevention, and other therapeutic applications.

Biochem/physiol Actions

Ectoine works as an enzyme stabilizer by maintaining hydration and reducing enzyme denaturation due to temperature changes. It also increases the stability of double-stranded DNA at high temperatures.

Other Notes

Stabilizer for enzymes and biological macromolecules

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Bownik et al.
Arhiv za higijenu rada i toksikologiju, 67(4), 260-265 (2016-12-30)
Ectoine is a compatible water molecule-binding solute (osmoprotectant) produced by several bacterial species in response to osmotic stress and unfavourable environmental conditions. This amino acid derivative can accumulate inside cells at high concentrations without interfering with natural processes and can
Ulrich Sydlik et al.
The European respiratory journal, 41(2), 433-442 (2012-10-27)
The life span of neutrophilic granulocytes has a determining impact on the intensity and duration of neutrophil driven lung inflammation. Based on the compatible solute ectoine, we aimed to prevent anti-apoptotic reactions in neutrophils triggered by the inflammatory microenvironment in
Alexander S Reshetnikov et al.
Extremophiles : life under extreme conditions, 15(6), 653-663 (2011-10-06)
The genes of ectoine biosynthesis pathway were identified in six species of aerobic, slightly halophilic bacteria utilizing methane, methanol or methylamine. Two types of ectoine gene cluster organization were revealed in the methylotrophs. The gene cluster ectABC coding for diaminobutyric
M.D. Vangala et al.
Bioforum, 23, 760-760 (2000)
Jens Smiatek et al.
Biophysical chemistry, 160(1), 62-68 (2011-10-22)
We have performed Molecular Dynamics simulations of ectoine, hydroxyectoine and urea in explicit solvent. Special attention has been spent on the local surrounding structure of water molecules. Our results indicate that ectoine and hydroxyectoine are able to accumulate more water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service