81280
Poly(ethylene glycol)
10,000
Synonym(s):
Polyethylene glycol, PEG
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
form
solid
mol wt
8500-11500
mp
62-65 °C
Ω-end
hydroxyl
α-end
hydroxyl
SMILES string
C(CO)O
InChI
1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
InChI key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Poly(ethylene glycol) (PEG) is an oligomer of ethylene oxide which is majorly used as a synthetic polymer. Its properties include hydrophilicity, biocompatibility, and low cost that make it useful in biomedical hydrogels.
Application
PEG can be used in a variety of applications ranging from drug delivery, biomedical applications and energy storage.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hydrogels in craniofacial tissue engineering
Biomaterials for Oral and Dental Tissue Engineering, 47-64 (2017)
Entrapment of essential oils in hydrogels for biomedical applications
Polymeric Gels, 125-141 (2018)
Biomedical hydrogels
Biomaterials, Artificial Organs and Tissue Engineering, 107-115 (2005)
Permeability and blood compatibility properties of chitosan-poly(ethylene oxide) blend membranes for haemodialysis
Amiji MM
Biomaterials, 16(8), 593-599 (1995)
Long-circulating poly (ethylene glycol)-modified gelatin nanoparticles for intracellular delivery
Kaul G and Amiji M
Pharmaceutical Research, 19(7), 1061-1067 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service