80757
Propamocarb-(propyl-d7)
PESTANAL®, analytical standard
Synonym(s):
Propamocarb-d7
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9D7H13N2O2
Molecular Weight:
195.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
Assay
≥98.0% (GC)
shelf life
limited shelf life, expiry date on the label
application(s)
agriculture
format
neat
storage temp.
2-8°C
SMILES string
CN(C)CCCNC(OC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O
InChI
1S/C9H20N2O2/c1-4-8-13-9(12)10-6-5-7-11(2)3/h4-8H2,1-3H3,(H,10,12)/i1D3,4D2,8D2
InChI key
WZZLDXDUQPOXNW-GZAMCDGTSA-N
Related Categories
General description
Propamocarb-(propyl-d7) is an isotope-labeled analog of systemic fungicide propamocarb, wherein propyl protons are substituted by deuterium.
Application
Propamocarb-(propyl-d7) may be used as a deuterated internal standard to quantify the pesticide analytes in foods of plant origin by two-dimensional liquid chromatography-tandem mass spectrometry (LC-MS/MS).
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.
Legal Information
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Quick method for the analysis of numerous highly polar pesticides in foods of plant origin via LC-MS/MS involving simultaneous extraction with methanol (QuPPe-method)
Anastassiades M, et al.
Chemosphere, 1117(2), 86-102 (2019)
Simultaneous determination of pesticides, mycotoxins, tropane alkaloids, growth regulators, and pyrrolizidine alkaloids in oats and whole wheat grains after online clean-up via two-dimensional liquid chromatography tandem mass spectrometry
Urban M, et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 54(2), 98-111 (2019)
Simultaneous determination of pesticides, mycotoxins, and metabolites as well as other contaminants in cereals by LC-LC-MS/MS
Kresse M, et al.
Journal of Chromatography. B, Biomedical Applications, 1117(2), 86-102 (2019)
Quick method for the analysis of numerous highly polar pesticides in foods of plant origin via LC-MS/MS involving simultaneous extraction with methanol (QuPPe-method)
Anastassiades M, et al.
EU Reference Laboratie for Residue of Pesticides, 9, 1-60 (2015)
Maurice Hiemstra et al.
Journal of chromatography. A, 972(2), 231-239 (2002-11-06)
A method for the determination of propamocarb in vegetables with liquid chromatography-electrospray ionisation mass spectrometry (LC-ESI-MS) was developed. The performance of a polymer-based analytical LC column for the separation was investigated. Residues of propamocarb were extracted from the matrix with
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service