Skip to Content
Merck
All Photos(1)

Key Documents

80443

SAFC

Sodium pyruvate

Pharma Manufacturing

Synonym(s):

α-Ketopropionic acid sodium salt, 2-Oxopropanoic acid sodium salt, Pyruvic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCOONa
CAS Number:
Molecular Weight:
110.04
Beilstein:
3568341
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

≥99.5% (HPLC)

form

solid

concentration

≥98 % (w/w) (T)

impurities

sodium, complies
≤0.2% every single impurity (HPLC)
≤0.5% water (Karl Fischer)
≤5000 ppm GC-HS: etahnol

mp

>300 °C (lit.)

solubility

H2O: soluble, clear, colorless (1.5g/25mL)

suitability

corresponds for identity (IR) (ATR)

SMILES string

[Na+].CC(=O)C([O-])=O

InChI

1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1

InChI key

DAEPDZWVDSPTHF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rekin's Janky et al.
PLoS computational biology, 10(7), e1003731-e1003731 (2014-07-25)
Identifying master regulators of biological processes and mapping their downstream gene networks are key challenges in systems biology. We developed a computational method, called iRegulon, to reverse-engineer the transcriptional regulatory network underlying a co-expressed gene set using cis-regulatory sequence analysis.
Tetsufumi Ito et al.
The Journal of comparative neurology, 523(15), 2277-2296 (2015-04-17)
Large GABAergic (LG) neurons form a distinct cell type in the inferior colliculus (IC), identified by the presence of dense VGLUT2-containing axosomatic terminals. Although some of the axosomatic terminals originate from local and commissural IC neurons, it has been unclear
Athanasia E Christakou et al.
Lab on a chip, 15(15), 3222-3231 (2015-07-02)
We demonstrate a simple method for three-dimensional (3D) cell culture controlled by ultrasonic standing waves in a multi-well microplate. The method gently arranges cells in a suspension into a single aggregate in each well of the microplate and, by this
Philipp Müller et al.
Cancer immunology research, 2(8), 741-755 (2014-06-12)
Antibody-drug conjugates (ADC) are emerging as powerful treatment strategies with outstanding target-specificity and high therapeutic activity in patients with cancer. Brentuximab vedotin represents a first-in-class ADC directed against CD30(+) malignancies. We hypothesized that its sustained clinical responses could be related
Philipp Müller et al.
Science translational medicine, 7(315), 315ra188-315ra188 (2015-11-27)
Targeted drug delivery with antibody-drug conjugates such as the HER2-directed ado-trastuzumab emtansine (T-DM1) has emerged as a powerful strategy for cancer therapy. We show that T-DM1 is particularly effective in eliciting antitumor immunity in patients with early breast cancer (WSG-ADAPT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service