Skip to Content
Merck
All Photos(2)

Key Documents

80073

Supelco

α-Hydroxymetoprolol

analytical standard

Synonym(s):

3-[4-(1-Hydroxy-2-methoxyethyl)phenoxy]-1-isopropylamino-2-propanol, 4-{2-Hydroxy-3-[(1-methylethyl)amino]propoxy}-α-(methoxymethyl)-benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H25NO4
CAS Number:
Molecular Weight:
283.36
Beilstein:
6571082
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

68-74 °C

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

OC(CNC(C)C)COC1=CC=C(C(O)COC)C=C1

InChI

1S/C15H25NO4/c1-11(2)16-8-13(17)9-20-14-6-4-12(5-7-14)15(18)10-19-3/h4-7,11,13,15-18H,8-10H2,1-3H3

InChI key

OFRYBPCSEMMZHR-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S W Ellis et al.
The Biochemical journal, 316 ( Pt 2), 647-654 (1996-06-01)
Cytochrome P-450 2D6 (CYP2D6) is an important human drug-metabolizing enzyme responsible for the oxidation of more than 30 widely used therapeutic agents. The enzymes encoded by the published genomic [Kimura, Umeno, Skoda, Meyer and Gonzalez (1989) Am. J. Hum. Genet.
Carrie A Morris et al.
Antimicrobial agents and chemotherapy, 58(10), 5900-5908 (2014-07-30)
The objectives of this study were to characterize any drug-drug interaction between the antimalarial Pyramax (pyronaridine-artesunate [PA]) and the CYP2D6 probe substrate metoprolol and to assess the safety of 60-day or 90-day PA redosing, particularly with regard to liver biochemistry
K Balmér et al.
Journal of chromatography, 553(1-2), 391-397 (1991-08-16)
The two enantiomers of metoprolol and the four enantiomeric forms of alpha-hydroxymetoprolol were separated by liquid chromatography on a Chiralcel OD column containing a cellulose tris(3,5-dimethyl-phenylcarbamate) chiral stationary phase. The column efficiency was strongly dependent on the flow-rate and the
H Shimizu et al.
Arzneimittel-Forschung, 42(6), 802-806 (1992-06-01)
The maximum plasma concentrations (Cmax) after oral administration of 120 mg tablets of slow-release metoprolol (CAS 37350-58-6) to 75 Japanese healthy male volunteers and 15 arrhythmic patients were measured. Extensive and poor metabolizers after oral administrations of slow-release metoprolol tablets
D R Sohn et al.
Therapeutic drug monitoring, 14(3), 184-189 (1992-06-01)
We examined the utility of the postdose 3-h plasma metabolic ratio (MR) as a phenotyping method for assessing genetically determined debrisoquine-type oxidation polymorphism after an oral dose of 100 mg of metoprolol tartrate administered to 402 unrelated, healthy, and native

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service