Skip to Content
Merck
All Photos(1)

Key Documents

793175

Sigma-Aldrich

Reagent Alcohol

ACS reagent

Synonym(s):

Laboratory Grade Ethanol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
15101802

grade

ACS reagent

Quality Level

vapor density

1.59 (vs air)

form

liquid

expl. lim.

3.1-27.7 % (lit.)

concentration

100%
94-96 % (v/v) (methanol SDA 3A 200 proof)

impurities

≤0.2% water
water

bp

78 °C (lit.)

mp

-114 °C (lit.)

Tax and license information

Sales restrictions may apply

Looking for similar products? Visit Product Comparison Guide

General description

Reagent alcohol is commonly used as a solvent in organic synthesis, analytical studies, and material synthesis.

Application

Useful for multiple research applications in universities and hospitals.

Packaging

4x1GA-PB is in poly bottle.
12x1GA-PB is 3x(4x1GA) in poly bottle.
20x1GA-PB is 5x(4x1GA) in poly bottle.
5GA is in metal drum with flex spout.
5GA-PB is in a poly drum with flex spout.
3x5GA is in metal drum with flex spout.
3x5GA-PB is in a poly drum with flex spout.
55GA is in metal drum.
55GA-PB is in poly drum.
270GA is in poly tote.

Components

95 parts of specially denatured ethyl alcohol 3A, 200 proof, ACS reagent, with 5 parts of isopropyl alcohol. Final concentrations are ~ 90% ethanol, ~ 5% methanol and ~ 5% isopropanol.

Other Notes

Ordering Information:
The letterhead approval is only required by Sigma-Aldrich when the unit size is 5GA or greater.

Link to Letterhead Example:

replacement part

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Controlled synthesis of graphene via electrochemical route and its use as efficient metal-free catalyst for oxygen reduction
Komba N, et al.
Applied Catalysis. B, Environmental, 243(9), 373-380 (2019)
A novel, density-independent and FTIR-compatible approach for the rapid extraction of microplastics from aquatic sediments
Crichton EM, et al.
Analytical Methods : Advancing Methods and Applications, 9(9), 1419-1428 (2017)
Design, synthesis and biological evaluation of benzamide and phenyltetrazole derivatives with amide and urea linkers as BCRP inhibitors
Gujarati NA, et al.
Bioorganic & Medicinal Chemistry Letters, 27(20), 4698-4704 (2017)
U Allenstein et al.
Soft matter, 11(25), 5053-5059 (2015-06-02)
Cells require adhesion to survive, proliferate and migrate, as well as for wound healing and many other functions. The strength of contractile cell forces on an underlying surface is a highly relevant quantity to measure the affinity of cells to
Nicholas J Niemuth et al.
Chemosphere, 135, 38-45 (2015-04-22)
The occurrence of intersex fish, where male reproductive tissues show evidence of feminization, have been found in freshwater systems around the world, indicating the potential for significant endocrine disruption across species in the ecosystem. Estrogens from birth control medications in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service