Skip to Content
Merck
All Photos(1)

Key Documents

78719

Millipore

Kovac′s Reagent Strips

suitable for microbiology

Synonym(s):

Indole Reagent Test sticks according to Kovac

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

sterility

sterile

Quality Level

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 25 strips

storage condition

(Prevent from moisture)

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

agriculture
clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Escherichia coli
enterobacteriaceae

General description

Kovac′s Reagent Strips are sterile filter paper strips impregnated with Kovac′s reagent used to detect indole-producing bacteria, primarily Enterobacteriaceae, to split indole from the tryptophan molecule. The various enzymes involved in the degradation of tryptophan to indole are collectively called tryptophanase, a general term used to denote the complete system of enzymes. The presence of indole is detected by Kovac′s reagent strip, which turns pink in the presence of indole.

Application

Kovac′s Reagent Strips are used to detect indole-producing bacteria, primarily Enterobacteriaceae species. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Packaging

package of 25 strips

Components

Sterile filter paper strips (diameter 7mm) impregnated with Kovac`s Reagent

Analysis Note

Escherichia coli (25922) - PositiveEnterobacter aerogenes (13048) - Negative

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satyajit Sarker
Chemistry for Pharmacy Students: General, Organic and Natural Product Chemistry (2013)

Articles

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service