77036
Manganese Standard for AAS
TraceCERT®, 1 g/L Mn in nitric acid (nominal concentration)
Synonym(s):
Manganese AAS Calibration Standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
concentration
1 g/L Mn in nitric acid (nominal concentration)
technique(s)
AAS: suitable
application(s)
industrial qc
pharmaceutical
format
single component solution
General description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Application
- Manganese AAS standard for environmental monitoring: Manganese standards for Atomic Absorption Spectroscopy (AAS) are essential in environmental monitoring, particularly in assessing manganese distribution in groundwater. This application is crucial for ensuring water safety and managing natural resources effectively (Rajmohan and Elango, 2005).
- Pharmaceutical manganese analysis: In pharmaceutical research, manganese AAS standards are used to ensure the precise quantification of manganese content in medicinal products. This is critical for meeting regulatory standards and ensuring patient safety (Cassella et al., 2011).
- Trace metal analysis manganese standard: High-purity manganese AAS standards are vital for trace metal analysis in various biotechnological applications. They help in the accurate measurement of manganese levels in biological samples, aiding in various research areas such as toxicology and nutrient metabolism (Borgese et al., 2011).
Analysis Note
1 g/L Mn in 2% nitric acid, prepared with high purity Mn metal, HNO3 and water
Other Notes
For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website
Legal Information
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service