Skip to Content
Merck
All Photos(1)

Key Documents

75041

Supelco

Neohesperidin dihydrochalcone

analytical standard

Synonym(s):

1-(4-((2-O-[6-Deoxy-α-L-mannopyranosyl]-β-D-glucopyranosyl)oxy)-2,6-dihydroxyphenyl)-3-[3-hydroxy-4-methoxyphenyl]-1-propanone, NHDC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H36O15
CAS Number:
Molecular Weight:
612.58
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤15% water

mp

156-158 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1ccc(CCC(=O)c2c(O)cc(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)cc2O)cc1O

InChI

1S/C28H36O15/c1-11-21(34)23(36)25(38)27(40-11)43-26-24(37)22(35)19(10-29)42-28(26)41-13-8-16(32)20(17(33)9-13)14(30)5-3-12-4-6-18(39-2)15(31)7-12/h4,6-9,11,19,21-29,31-38H,3,5,10H2,1-2H3/t11-,19+,21-,22+,23+,24-,25+,26+,27-,28+/m0/s1

InChI key

ITVGXXMINPYUHD-CUVHLRMHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Neohesperidin dihydrochalcone is a non-nutritive sweetening agent, widely used as flavor enhancer and flavor modifier in food products. It is also involved in improving the sweetness profile and suppressing bitterness/saltiness properties.

Application

Neohesperidin dihydrochalcone may be used as an analytical reference standard for the quantification of the analyte in fruits and wastewater samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of eight artificial sweeteners in wastewater by hydrophilic interaction liquid chromatography-tandem mass spectrometry
Kokotou.GM and Thomaidis.SN
Analytical Methods : Advancing Methods and Applications, 5(16), 3825-3833 (2016)
Food Analysis by HPLC, Second Edition, 100(2) (2000)
Nollet .L.ML
Food Analysis by HPLC, Second Edition, (2) (2000)
Hesperidin and neohesperidin dihydrochalcone on different experimental models of induced gastric ulcer
Suarez.J, et al.
Phytotherapy Research, 10(7), 616- 618 (1996)
A high-pressure liquid chromatographic method for the quantitation of neohesperidin dihydrochalcone
Fisher.FJ
Journal of Agricultural and Food Chemistry, 25(3), 682-683 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service