Skip to Content
Merck
All Photos(1)

Key Documents

74316

Supelco

Sodium 1-nonanesulfonate

suitable for ion pair chromatography, LiChropur, ≥99.0% (T)

Synonym(s):

1-Nonanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8SO3Na
CAS Number:
Molecular Weight:
230.30
Beilstein:
3738676
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

anionic

Quality Level

Assay

≥99.0% (T)

form

powder

quality

LiChropur

mol wt

230.30 g/mol

technique(s)

ion pair chromatography: suitable

mp

>300 °C (lit.)

λ

5 % in H2O

UV absorption

λ: 210 nm Amax: 0.05
λ: 220 nm Amax: 0.03
λ: 230 nm Amax: 0.02
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

[Na+].CCCCCCCCCS([O-])(=O)=O

InChI

1S/C9H20O3S.Na/c1-2-3-4-5-6-7-8-9-13(10,11)12;/h2-9H2,1H3,(H,10,11,12);/q;+1/p-1

InChI key

RUYRDULZOKULPK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium 1-nonanesulfonate is an ion pair reagent, suitable for cation separation sorted by carbon chain length.

Application

Sodium 1-nonanesulfonate may be used as a mobile phase additive for the determination of cefmatilen and its metabolites in biological samples by coupled systems of ion-exchange and reversed-phase high-performance liquid chromatography.
Sodium 1-nonanesulfonate may also be used as an ion-pair reagent in liquid chromatography analysis of guanidino compounds in serum using a porous graphite carbon column.

Other Notes

Ion-pair reagent used in the separation of histidines by HPLC

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid analysis of 3-methylhistidine in urine, plasma, muscle and amniotic fluid with a single high-performance liquid chromatographic system but with different ion-pairing reagents.
W S Hancock et al.
Journal of chromatography, 228, 273-278 (1982-03-12)
Determination of a new oral cephalosporin, cefmatilen hydrochloride hydrate, and its seven metabolites in human and animal plasma and urine by coupled systems of ion-exchange and reversed-phase high-performance liquid chromatography
Nishino I, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 749(1), 101-110 (2000)
Transport properties for aqueous sodium sulfonate surfactants: 2. Intradiffusion measurements: influence of the obstruction effect on the monomer and micelle mobilities
Annunziata O, et al.
Journal of Colloid and Interface Science, 216(1), 16-24 (1999)
Liquid chromatography of guanidino compounds using a porous graphite carbon column and application to their analysis in serum
Inamoto Y, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 707(1-2), 111-120 (1998)
Synthesis, characterization and reaction behaviour of lamellar AFm phases with aliphatic sulfonate-anions
Pollmann H, et al.
Cement and Concrete Research, 36(11), 2039-2048 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service