Skip to Content
Merck
All Photos(2)

Key Documents

74158

Supelco

Nonadecane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17CH3
CAS Number:
Molecular Weight:
268.52
Beilstein:
1742892
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

9.27 (vs air)

vapor pressure

1 mmHg ( 133.2 °C)

Assay

≥99.5% (GC)

autoignition temp.

446 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

330 °C (lit.)

mp

30-34 °C (lit.)
32-34 °C

density

0.786 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCC

InChI

1S/C19H40/c1-3-5-7-9-11-13-15-17-19-18-16-14-12-10-8-6-4-2/h3-19H2,1-2H3

InChI key

LQERIDTXQFOHKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefano Colazza et al.
Journal of chemical ecology, 33(7), 1405-1420 (2007-05-18)
Contact kairomones from adult southern green stink bugs, Nezara viridula (L.) (Heteroptera: Pentatomidae) that elicit foraging behavior of the egg parasitoid Trissolcus basalis (Wollaston) were investigated in laboratory experiments. Chemical residues from tarsi and scutella of N. viridula induced foraging
Abdiwahab A Musse et al.
The Journal of biological chemistry, 281(2), 885-895 (2005-11-22)
Helix 1 of the membrane-associated closed state of the colicin E1 channel domain was studied by site-directed fluorescence labeling where bimane was covalently attached to a single cysteine residue in each mutant protein. A number of fluorescence properties of the
Daniela Lo Giudice et al.
Journal of chemical ecology, 37(6), 622-628 (2011-05-07)
Egg parasitoids are able to find their hosts by exploiting their chemical footprints as host location cues. In nature, the apolar epicuticular wax layer of plants that consists of several classes of hydrocarbons serves as the substrate that retains these
Stefano Colazza et al.
Die Naturwissenschaften, 96(8), 975-981 (2009-05-21)
Chemical footprints left behind by true bugs are perceived as contact kairomones by scelionid egg parasitoids. Female wasps encountering a contaminated artificial substrate display a characteristic arrestment posture, holding the body motionless and antennating the surface. In the system Nezara
C Pidgeon et al.
Analytical biochemistry, 181(1), 28-32 (1989-08-15)
Quantitating the lipid content in organic lipid solutions and extracted membrane preparations is described. Fourier transform infrared analysis of thin films using perdeuterated nonadecane as an internal standard permitted quantitation with greater than 95% accuracy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service