Skip to Content
Merck
All Photos(1)

Key Documents

73664

Supelco

Mettler-Toledo Calibration substance ME 18870, Benzophenone

traceable to primary standards (LGC)

Synonym(s):

Benzophenone, Diphenyl ketone, Diphenylmethanone, NSC 8077

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CO
CAS Number:
Molecular Weight:
182.22
Beilstein:
1238185
EC Number:
MDL number:
UNSPSC Code:
20121904
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
for the calibration of the thermosystem 900

Quality Level

vapor pressure

1 mmHg ( 108 °C)

quality

traceable to primary standards (LGC)

shelf life

limited shelf life, expiry date on the certificate and label

bp

305 °C (lit.)

mp

47-51 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

SMILES string

O=C(C1=CC=CC=C1)C2=CC=CC=C2

InChI

1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

RWCCWEUUXYIKHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mettler-Toledo calibration substance ME 18870, benzophenone (47-51 °C) is an analytical standard, ideal for the routine calibration of Mettler-Toledo melting point instrument. The melting point valueequals a mean of 6 to 12 measurements with aMettler-Toledo MP90 Excellence instrument that is calibrated withprimary standards.

Application

This Mettler-Toledo melting point standard is used to calibrate the melting point instrument Mettler FP900 thermosystem.

Features and Benefits

  • Melting point calibration standard traceable to a primary standard from LGC, London
  • Melting point evaluation performed in both thermodynamic and pharmacopeia modes for physically correct and heating rate dependent melting point determinations, respectively
  • Available with certificates of analysis and safety data sheet
  • A product of analytical standard grade to help meet the QC/QA requirements of melting point determination

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - STOT RE 2 Oral

Target Organs

Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erythritol: crystal growth from the melt
Jesus Lopes.J.A, et al.
International Journal of Pharmaceutics, 388(1), 129-135 (2010)
Masayuki Kyomoto et al.
Biomaterials, 34(32), 7829-7839 (2013-07-31)
We investigated the production of free radicals on a poly(ether-ether-ketone) (PEEK) substrate under ultraviolet (UV) irradiation. The amount of the ketyl radicals produced from the benzophenone (BP) units in the PEEK molecular structure initially increased rapidly and then became almost
Young-Jin Kim et al.
Angewandte Chemie (International ed. in English), 51(42), 10537-10541 (2012-09-20)
Caught in a web: Photo-cross-linkable temperature-responsive polymer-based nanofiber webs have been synthesized that have the ability to capture, encapsulate, and release cells by dynamically transforming the fibrous structure into hydrogel-like structures by wrapping, swelling, and deswelling processes in response to
L L Barnkob et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 30(2), 395-402 (2012-11-17)
The aim of this study was to investigate the effect of relative humidity on the migration of benzophenone from paperboard into the food simulant Tenax®. Kinetic migration investigations were carried out with three relative humidities in the interval between 39%
Haidong Li et al.
Talanta, 99, 811-815 (2012-09-13)
Benzophenone is one of the most commonly used photoinitiators of UV-cured inks on food packaging materials and can migrate into foodstuffs. In this study, an amperometric benzophenone sensor based on molecularly imprinted polymer (MIP) was successfully constructed for the first

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service