Skip to Content
Merck
All Photos(1)

Key Documents

73137

Supelco

2,2-Dimethoxypropane

analytical standard

Synonym(s):

Acetone dimethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(OCH3)2
CAS Number:
Molecular Weight:
104.15
Beilstein:
635678
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.02

grade

analytical standard

Quality Level

vapor density

3.59 (vs air)

vapor pressure

60 mmHg ( 15.8 °C)

Assay

≥99.4% (GC)

shelf life

limited shelf life, expiry date on the label

expl. lim.

31 %, 58 °F
6 %, 27 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.378 (lit.)
n20/D 1.378

bp

79-81 °C
83 °C (lit.)

density

0.847 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

COC(C)(C)OC

InChI

1S/C5H12O2/c1-5(2,6-3)7-4/h1-4H3

InChI key

HEWZVZIVELJPQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2-Dimethoxypropane is an organic compound, which may be used as a protecting agent in the process of the synthesis of structurally related alkaloids like narciclasine and lycoricidine using phenylbromide as the starting material.

Application

2,2-Dimethoxypropane may be used as an analytical standard for the determination of the analyte in biological samples by headspace gas chromatography-mass spectrometry (HS-GC/MS) technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of n-propane, iso-butane, and n-butane by headspace GC-MS in intoxications by inhalation of lighter fluid
Bouche MPLA, et al.
Journal of Analytical Toxicology, 26(1), 35-42 (2002)
Total Synthesis of Lycoricidine and Narciclasine by Chemical Dearomatization of Bromobenzene
Southgate.HE, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 129(47), 15245-15248 (2017)
H J Beckmann et al.
Histochemistry, 76(3), 407-412 (1982-01-01)
Dehydration with 2,2-dimethoxypropane acidified with either 1 N HCL or 0.05 N HCL in an 1:100 ratio has been found to extract various biologically active lipids. The effect is enhanced by an increased proton concentration and seems to occur only
Akito Takeuchi et al.
Biomedical chromatography : BMC, 24(5), 465-471 (2009-08-19)
A method for routinely determination of dimethyl sulfoxide (DMSO) and dimethyl sulfone (DMSO(2)) in human urine was developed using gas chromatography-mass spectrometry. The urine sample was treated with 2,2-dimethoxypropane (DMP) and hydrochloric acid for efficient removal of water, which causes
[Quantitative double-labeled autoradiography by chemical washing method using 2,2-dimethoxypropane].
H Sumiya et al.
Kaku igaku. The Japanese journal of nuclear medicine, 24(3), 315-318 (1987-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service