Skip to Content
Merck
All Photos(1)

Key Documents

72405

Supelco

1,2-Dimethoxyethane

analytical standard

Synonym(s):

mono-Glyme, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OCH3
CAS Number:
Molecular Weight:
90.12
Beilstein:
1209237
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.1 (20 °C, vs air)

vapor pressure

48 mmHg ( 20 °C)

Assay

≥99.7% (GC)

autoignition temp.

396 °F

shelf life

limited shelf life, expiry date on the label

contains

~0.01% 2,6-di-tert-butyl-4-methylphenol as stabilizer

expl. lim.

10.4 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.379 (lit.)
n20/D 1.380

bp

85 °C (lit.)

mp

−58 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

COCCOC

InChI

1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3

InChI key

XTHFKEDIFFGKHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dimethoxyethane is a water miscible solvent, used as an alternative for the hazardous organic solvents such as dioxane and tetrahydrofuran.

Application

1,2-Dimethoxyethane may be used as a solvent for the separation and determination of neutral sugars as 2,4- dinitrophenyl derivatives using high-performance liquid chromatography.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

recommended

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Analysis of neutral sugars as dinitrophenyl-hydrazones by high-performance liquid chromatography
Karamanos.K.N, et al.
Journal of Chromatography A, 405, 221-228 (1987)
Introduction to Organic Laboratory Techniques: A Small Scale Approach (2005)
Shaokun Tang et al.
Bioresource technology, 129, 667-671 (2013-01-10)
Glymes (i.e. glycol diethers) were explored as alternative benign solvents for enzymatic reactions, specifically the lipase-catalyzed transesterification. Long-chain glymes were found highly compatible with immobilized Candida antarctica lipase B (iCALB), leading to higher enzyme activities and stabilities than t-butanol and
Andrew Martins et al.
Organic letters, 12(22), 5186-5188 (2010-10-19)
A palladium-catalyzed crossed biaryl coupling/reduction sequence enables the formation of meta-substituted biaryls via solvent-mediated arylpalladium(II) reduction. Isotope labeling studies determined that the decomposition of 1,2-dimethoxyethane (DME) is indeed involved in the reductive process.
Jenn-Huei Lii et al.
Journal of computational chemistry, 24(12), 1490-1503 (2003-07-18)
Ethylene glycol, its dimethyl ether, and some related compounds have been studied using the MM4 molecular mechanics force field. The MM4 calculated structural and energetic results have been brought into satisfactory agreement with a considerable number of experimental data and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service