Skip to Content
Merck
All Photos(1)

Key Documents

71732

Supelco

Sodium ionophore I

Selectophore, function tested

Synonym(s):

ETH 227, N,N′,N′′-Triheptyl-N,N′,N′′-trimethyl-4,4′,4′′-propylidynetris(3-oxabutyramide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H71N3O6
CAS Number:
Molecular Weight:
641.97
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

liquid

quality

function tested

SMILES string

CCCCCCCN(C)C(=O)COCC(CC)(COCC(=O)N(C)CCCCCCC)COCC(=O)N(C)CCCCCCC

InChI

1S/C36H71N3O6/c1-8-12-15-18-21-24-37(5)33(40)27-43-30-36(11-4,31-44-28-34(41)38(6)25-22-19-16-13-9-2)32-45-29-35(42)39(7)26-23-20-17-14-10-3/h8-32H2,1-7H3

InChI key

PMECOBBGCPFYPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Other Notes

Neutral ionophore for liquid-membrane electrodes of high selectivity for Na+:; R.A. Steiner, M. Oehme, D. Ammann, W. Simon,.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maheswari Jayakannan et al.
Journal of plant physiology, 168(10), 1045-1051 (2011-01-25)
Ion-selective microelectrodes are a powerful tool in studying adaptive responses of plant cells and tissues to various abiotic stresses. However, application of this technique in Na(+) flux measurements was limited due to poor selectivity for Na(+) ions of commercially available
Z Talor et al.
Mineral and electrolyte metabolism, 12(4), 239-245 (1986-01-01)
We investigated the Na-dependent Ca transport in purified bovine luminal and basolateral renal tubular membranes. Na-dependent Ca uptake was observed in basolateral but not in luminal kidney tubular membranes. 45Ca uptake in basolateral membrane vesicles loaded with Na and suspended
D E Carden et al.
Journal of experimental botany, 52(359), 1353-1359 (2001-07-04)
The high background K+ concentration in plant cells is a problem for intracellular measurements of Na+ using ion-selective microelectrodes. The discrimination between Na+ and K+ of the microelectrode ionophore molecule limits the usefulness of this technique. A new Na+-selective microelectrode
A Amtmann et al.
The Journal of membrane biology, 139(2), 117-125 (1994-04-01)
Na(+)-selective microelectrodes with the sensor ETH 227 have been used to measure the cytoplasmic Na+ concentration, [Na+]c, in Acetabularia. In the steady-state, [Na+]c is about 60 mM (external 460 mM). Steps in external Na+ concentration, [Na+]o, cause biexponential relaxations of
Analytical Chemistry, 51, 351-351 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service